Search
Search

SLC2A5| Gene Symbol: SLC2A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003039.2 | NP_003030.1 | 12 | 2021 | ||
| XM_005263491.3 | XP_005263548.1 | 13 | 1819 | |||
| XM_017002133.1 | XP_016857622.1 | 14 | 2047 | |||
| XM_017002134.1 | XP_016857623.1 | 14 | 2013 | |||
| XM_017002135.1 | XP_016857624.1 | 14 | 2107 | |||
| XM_017002136.1 | XP_016857625.1 | 14 | 2083 | |||
| XM_017002137.1 | XP_016857626.1 | 15 | 2119 | |||
| XM_017002138.1 | XP_016857627.1 | 15 | 2095 | |||
| XM_017002139.1 | XP_016857628.1 | 13 | 2029 | |||
| XM_017002140.1 | XP_016857629.1 | 14 | 1950 | |||
| XM_017002141.1 | XP_016857630.1 | 13 | 1852 | |||
| XM_017002142.1 | XP_016857631.1 | 10 | 1592 | |||
| GenBank mRNA | AK294368.1 | - | 12 | 1807 | ||
| AK300477.1 | - | 10 | 1597 | |||
| AK302057.1 | - | 15 | 1945 | |||
| AK302137.1 | - | 14 | 1897 | |||
| BC001692.2 | - | 12 | 1780 | 2966459 | ||
| BC001820.2 | - | 12 | 1780 | 2966459 | ||
| M55531.1 | - | 12 | 1804 | |||