Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST3GAL5 SIRNA Assays ›
Gene Symbol:
ST3GAL5
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Gene Aliases:
SATI, SIAT9, SIATGM3S, ST3GalV
Chromosome Location:
Chr.2: 85839148 - 85889034 on Build GRCh38
UniGene ID:
Hs.415117
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
117201
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST3GAL5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042437.1
NP_001035902.1
4
634
NM_003896.3
NP_003887.3
4
779
XM_005264630.4
XP_005264687.1
4
735
XM_011533143.2
XP_011531445.1
4
907
XM_017005199.1
XP_016860688.1
5
957
XM_017005200.1
XP_016860689.1
5
1851
XM_017005201.1
XP_016860690.1
4
742
XM_017005202.1
XP_016860691.1
5
917
XM_017005203.1
XP_016860692.1
6
1415
XM_017005204.1
XP_016860693.1
7
1496
XM_017005205.1
XP_016860694.1
7
1515
XM_017005206.1
XP_016860695.1
6
1293
XM_017005207.1
XP_016860696.1
5
893
XM_017005208.1
XP_016860697.1
7
1418
XM_017005209.1
XP_016860698.1
6
1075
XM_017005210.1
XP_016860699.1
6
2422
XM_017005211.1
XP_016860700.1
5
1067
XM_017005212.1
XP_016860701.1
5
1805
XM_017005213.1
XP_016860702.1
2
529
XM_017005214.1
XP_016860703.1
4
907
XM_017005215.1
XP_016860704.1
5
1014
GenBank mRNA
AB018356.1
-
5
759
AF105026.1
-
4
713
AF119415.1
-
5
678
AF119417.1
-
1
1704
AF119418.1
-
4
964
AK001340.1
-
4
736
AK127346.1
-
2
2999
AY152815.2
-
4
634
AY359105.1
-
4
639
BC065936.1
-
5
695
6163586
KU178447.1
-
3
566
KU178448.1
-
3
566
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
carbohydrate metabolic process
glycosphingolipid biosynthetic process
oligosaccharide metabolic process
protein N-linked glycosylation via asparagine
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
neolactotetraosylceramide alpha-2,3-sialyltransferase activity
sialyltransferase activity
lactosylceramide alpha-2,3-sialyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0