Search
Search

OVGP1| Gene Symbol: OVGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002557.3 | NP_002548.3 | 3 | 280 | ||
| GenBank mRNA | AB208783.1 | - | 1 | 596 | ||
| AK295207.1 | - | 2 | 620 | |||
| AK298757.1 | - | 2 | 208 | |||
| AK314074.1 | - | 3 | 238 | |||
| AY189737.1 | - | 3 | 281 | |||
| BC126177.1 | - | 3 | 263 | 8991893 | ||
| BC136406.1 | - | 3 | 263 | 9020393 | ||
| U09550.1 | - | 3 | 236 | |||