Search
Search

OVGP1| Gene Symbol: OVGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002557.3 | NP_002548.3 | 8 | 947 | ||
| GenBank mRNA | AB208783.1 | - | 6 | 1329 | ||
| AK298757.1 | - | 6 | 690 | |||
| AK314074.1 | - | 8 | 905 | |||
| AY189737.1 | - | 8 | 948 | |||
| BC126177.1 | - | 8 | 930 | 8991893 | ||
| BC136406.1 | - | 8 | 930 | 9020393 | ||
| U09550.1 | - | 8 | 903 | |||