Search
Search

KIF2B| Gene Symbol: KIF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032559.4 | NP_115948.4 | 1 | 354 | ||
| GenBank mRNA | AF333335.2 | - | 1 | 332 | ||
| AK057272.1 | - | 1 | 331 | |||
| BC033802.1 | - | 1 | 292 | 5170969 | ||
| DB457485.1 | - | 1 | 354 | |||