Search
Search

KIF2B| Gene Symbol: KIF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032559.4 | NP_115948.4 | 1 | 2057 | ||
| GenBank mRNA | AF333335.2 | - | 1 | 2035 | ||
| AK057272.1 | - | 1 | 2034 | |||
| AK302262.1 | - | 3 | 1653 | |||
| AK308265.1 | - | 2 | 1636 | |||
| BC033802.1 | - | 1 | 1995 | 5170969 | ||