Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDXDC1 SIRNA Assays ›
Gene Symbol:
PDXDC1
Gene Name:
pyridoxal dependent decarboxylase domain containing 1
Gene Aliases:
LP8165
Chromosome Location:
Chr.16: 14974591 - 15139410 on Build GRCh38
UniGene ID:
Hs.370781
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118784
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDXDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001285444.1
NP_001272373.1
15, 16
1560
NM_001285445.1
NP_001272374.1
15, 16
1557
NM_001285447.1
NP_001272376.1
16, 17
1453
NM_001285448.1
NP_001272377.1
15, 16
1466
NM_001324019.1
NP_001310948.1
16, 17
1638
NM_015027.3
NP_055842.2
16, 17
1641
XM_005255173.1
XP_005255230.1
16, 17
1597
XM_005255176.2
XP_005255233.1
16, 17
1523
XM_006720865.2
XP_006720928.2
15, 16
1359
XM_017023059.1
XP_016878548.1
17, 18
1533
XM_017023060.1
XP_016878549.1
16, 17
1703
XM_017023061.1
XP_016878550.1
16, 17
1473
XM_017023062.1
XP_016878551.1
16, 17
1554
XM_017023063.1
XP_016878552.1
17, 18
1533
XM_017023064.1
XP_016878553.1
15, 16
1461
GenBank mRNA
AK025504.1
-
3, 4
275
AK295168.1
-
16, 17
1446
AK299111.1
-
15, 16
1391
AK299799.1
-
15, 16
1451
BC025366.2
-
10, 11
827
BC036520.1
-
16, 17
1505
5270048
BC060871.1
-
16, 17
1475
30347972
BX647809.1
-
15, 16
1401
BX648066.1
-
15, 16
1398
D87438.1
-
16, 17
1501
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
carboxylic acid metabolic process
cell-cell adhesion
carboxy-lyase activity
pyridoxal phosphate binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0