Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CADPS SIRNA Assays ›
          Gene Symbol:
          CADPS
          Gene Name:
          calcium dependent secretion activator
          Gene Aliases:
          CADPS1, CAPS, CAPS1, UNC-31
          Chromosome Location:
          Chr.3: 62398346 - 62875389 on Build GRCh38
          UniGene ID:
          Hs.654933
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 118976

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CADPS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003716.3 NP_003707.2 7, 8 1793
          NM_183393.2 NP_899630.1 7, 8 1793
          NM_183394.2 NP_899631.1 7, 8 1793
          XM_006713378.2 XP_006713441.2 7, 8 1788
          XM_011534177.1 XP_011532479.1 7, 8 1788
          XM_011534178.1 XP_011532480.1 7, 8 1788
          XM_011534180.1 XP_011532482.1 7, 8 1788
          XM_011534182.1 XP_011532484.1 7, 8 1788
          XM_011534183.1 XP_011532485.1 7, 8 1788
          XM_011534189.1 XP_011532491.1 7, 8 1788
          XM_011534191.2 XP_011532493.1 7, 8 1433
          XM_011534192.1 XP_011532494.1 7, 8 1788
          XM_011534193.1 XP_011532495.1 7, 8 1788
          XM_011534194.2 XP_011532496.1 7, 8 1433
          XM_011534195.1 XP_011532497.1 7, 8 1788
          XM_011534196.1 XP_011532498.1 7, 8 1788
          XM_011534197.2 XP_011532499.1 7, 8 1746
          XM_011534198.2 XP_011532500.1 7, 8 1433
          XM_011534199.2 XP_011532501.1 7, 8 1433
          XM_011534200.2 XP_011532502.1 7, 8 1433
          XM_011534202.2 XP_011532504.1 7, 8 1788
          XM_011534203.1 XP_011532505.1 7, 8 1788
          XM_017007358.1 XP_016862847.1 7, 8 1788
          XM_017007359.1 XP_016862848.1 7, 8 1788
          XM_017007360.1 XP_016862849.1 7, 8 1788
          XM_017007361.1 XP_016862850.1 7, 8 1788
          XM_017007362.1 XP_016862851.1 7, 8 1788
          XM_017007363.1 XP_016862852.1 7, 8 1788
          XM_017007364.1 XP_016862853.1 7, 8 1788
          XM_017007365.1 XP_016862854.1 7, 8 1788
          XM_017007366.1 XP_016862855.1 7, 8 1788
          XM_017007367.1 XP_016862856.1 7, 8 1433
          XM_017007368.1 XP_016862857.1 7, 8 1433
          XM_017007369.1 XP_016862858.1 7, 8 1788
          XM_017007370.1 XP_016862859.1 7, 8 1788
          XM_017007371.1 XP_016862860.1 7, 8 1788
          XM_017007372.1 XP_016862861.1 7, 8 1433
          XM_017007373.1 XP_016862862.1 7, 8 1788
          XM_017007374.1 XP_016862863.1 7, 8 1788
          XM_017007375.1 XP_016862864.1 7, 8 1433
          XM_017007376.1 XP_016862865.1 7, 8 1788
          XM_017007377.1 XP_016862866.1 7, 8 1433
          XM_017007378.1 XP_016862867.1 7, 8 1433
          XM_017007379.1 XP_016862868.1 7, 8 1788
          XM_017007380.1 XP_016862869.1 7, 8 1788
          XM_017007381.1 XP_016862870.1 7, 8 1788
          XM_017007382.1 XP_016862871.1 7, 8 1433
          XM_017007383.1 XP_016862872.1 7, 8 1788
          XM_017007384.1 XP_016862873.1 7, 8 1788
          XM_017007385.1 XP_016862874.1 7, 8 1788
          XM_017007386.1 XP_016862875.1 7, 8 1788
          GenBank mRNA AB032947.2 - 7, 8 1783
          AB593079.1 - 7, 8 1746
          AF458662.1 - 7, 8 1743
          AL833895.1 - 7, 8 1090

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          protein transport
          vesicle organization
          synaptic vesicle priming
          catecholamine secretion
          dense core granule exocytosis
          protein binding
          lipid binding
          protein kinase binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0