Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF7 SIRNA Assays ›
Gene Symbol:
ARHGEF7
Gene Name:
Rho guanine nucleotide exchange factor 7
Gene Aliases:
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
Chromosome Location:
Chr.13: 111114619 - 111305734 on Build GRCh38
UniGene ID:
Hs.508738
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119396
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113511.2
NP_001106983.1
8
1803
NM_001113512.2
NP_001106984.1
6
1653
NM_001113513.1
NP_001106985.1
6
745
NM_001320851.1
NP_001307780.1
5
526
NM_001320852.1
NP_001307781.1
7
1740
NM_001320853.1
NP_001307782.1
6
651
NM_001320854.1
NP_001307783.1
2
265
NM_003899.3
NP_003890.1
6
858
NM_145735.3
NP_663788.1
7
1740
XM_006719956.3
XP_006720019.1
7
868
XM_011521133.2
XP_011519435.1
7
868
XM_017020814.1
XP_016876303.1
7
868
XM_017020815.1
XP_016876304.1
5
861
XM_017020816.1
XP_016876305.1
6
636
XM_017020817.1
XP_016876306.1
5
861
XM_017020818.1
XP_016876307.1
7
736
XM_017020819.1
XP_016876308.1
6
745
XM_017020820.1
XP_016876309.1
6
868
XM_017020821.1
XP_016876310.1
6
563
XM_017020822.1
XP_016876311.1
5
559
XM_017020823.1
XP_016876312.1
5
1336
XM_017020824.1
XP_016876313.1
7
736
XM_017020825.1
XP_016876314.1
5
487
XM_017020826.1
XP_016876315.1
7
736
XM_017020827.1
XP_016876316.1
5
560
XM_017020828.1
XP_016876317.1
2
125
XM_017020829.1
XP_016876318.1
2
125
GenBank mRNA
AB177848.1
-
7
1741
AB177849.1
-
4
464
AJ844629.1
-
3
188
AK295472.1
-
2
190
AK300272.1
-
6
638
AK308897.1
-
2
265
AL834228.1
-
7
924
BC033905.1
-
6
850
5262912
BC050521.1
-
6
717
BC060776.1
-
8
1145
5271977
D63476.1
-
6
834
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
Golgi organization
signal transduction
nervous system development
positive regulation of fibroblast migration
lamellipodium assembly
positive regulation of protein binding
regulation of Rho protein signal transduction
intracellular signal transduction
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of apoptotic process
positive regulation of GTPase activity
ephrin receptor signaling pathway
focal adhesion assembly
regulation of small GTPase mediated signal transduction
positive regulation of growth hormone secretion
positive regulation of substrate adhesion-dependent cell spreading
regulation of GTP binding
positive regulation of lamellipodium morphogenesis
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0