Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP9B SIRNA Assays ›
Gene Symbol:
ATP9B
Gene Name:
ATPase phospholipid transporting 9B (putative)
Gene Aliases:
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
Chromosome Location:
Chr.18: 79069275 - 79378391 on Build GRCh38
UniGene ID:
Hs.465475
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
120781
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP9B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306085.1
NP_001293014.1
4
661
NM_198531.4
NP_940933.3
4
661
XM_011525963.2
XP_011524265.1
5
827
XM_011525964.2
XP_011524266.1
4
542
XM_011525965.2
XP_011524267.1
5
823
XM_011525966.2
XP_011524268.1
4
778
XM_017025726.1
XP_016881215.1
5
827
XM_017025727.1
XP_016881216.1
5
827
XM_017025728.1
XP_016881217.1
5
827
XM_017025729.1
XP_016881218.1
5
827
XM_017025730.1
XP_016881219.1
5
827
XM_017025731.1
XP_016881220.1
5
827
XM_017025732.1
XP_016881221.1
5
827
XM_017025733.1
XP_016881222.1
5
827
XM_017025734.1
XP_016881223.1
5
827
XM_017025735.1
XP_016881224.1
5
827
XM_017025736.1
XP_016881225.1
5
827
XM_017025737.1
XP_016881226.1
5
827
XM_017025738.1
XP_016881227.1
2
232
XM_017025739.1
XP_016881228.1
2
232
XM_017025740.1
XP_016881229.1
2
232
XM_017025741.1
XP_016881230.1
2
232
XM_017025742.1
XP_016881231.1
5
827
GenBank mRNA
AB373110.1
-
3
369
AK093704.1
-
3
356
AK097757.1
-
4
778
AK295981.1
-
4
528
AK308291.1
-
4
543
BC053561.1
-
4
446
BC125219.1
-
4
539
40123958
BC125220.1
-
4
539
40123959
BX537860.1
-
1
12
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
retrograde vesicle-mediated transport, Golgi to ER
endocytosis
phospholipid translocation
establishment of protein localization to Golgi
magnesium ion binding
phospholipid-translocating ATPase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0