Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNC SIRNA Assays ›
          Gene Symbol:
          TNC
          Gene Name:
          tenascin C
          Gene Aliases:
          150-225, DFNA56, GMEM, GP, HXB, JI, TN, TN-C
          Chromosome Location:
          Chr.9: 115019575 - 115118257 on Build GRCh38
          UniGene ID:
          Hs.143250
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 121365

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002160.3 NP_002151.2 3 1791
          XM_005251972.3 XP_005252029.1 3 1741
          XM_005251973.3 XP_005252030.1 3 1741
          XM_005251974.3 XP_005252031.1 3 1741
          XM_005251975.3 XP_005252032.1 3 1741
          XM_006717096.3 XP_006717159.1 3 1741
          XM_006717097.3 XP_006717160.1 3 1741
          XM_006717098.3 XP_006717161.1 3 1741
          XM_006717101.3 XP_006717164.1 3 1741
          XM_011518625.2 XP_011516927.1 3 1741
          XM_011518626.2 XP_011516928.1 3 1741
          XM_011518628.2 XP_011516930.1 3 1741
          XM_011518629.2 XP_011516931.1 3 1741
          XM_017014678.1 XP_016870167.1 3 1741
          XM_017014679.1 XP_016870168.1 3 1741
          XM_017014680.1 XP_016870169.1 3 1741
          XM_017014681.1 XP_016870170.1 3 1741
          GenBank mRNA AB210038.1 - 3 1667
          AK304015.1 - 3 1687
          BC151843.1 - 3 1659 8860362
          GU473235.1 - 2 1379
          M24630.1 - 1 89
          M55618.1 - 2 1514
          X56160.1 - 2 1430
          X78565.1 - 3 1692

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          cell adhesion
          negative regulation of cell adhesion
          neuromuscular junction development
          positive regulation of cell proliferation
          response to wounding
          response to mechanical stimulus
          positive regulation of gene expression
          peripheral nervous system axon regeneration
          extracellular matrix organization
          wound healing
          odontogenesis of dentin-containing tooth
          response to ethanol
          bud outgrowth involved in lung branching
          mesenchymal-epithelial cell signaling involved in prostate gland development
          prostate gland epithelium morphogenesis
          cellular response to retinoic acid
          cellular response to vitamin D
          response to fibroblast growth factor
          cellular response to prostaglandin D stimulus
          syndecan binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0