Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LIF SIRNA Assays ›
Gene Symbol:
LIF
Gene Name:
leukemia inhibitory factor
Gene Aliases:
CDF, DIA, HILDA, MLPLI
Chromosome Location:
Chr.22: 30240447 - 30246851 on Build GRCh38
UniGene ID:
Hs.2250
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121369
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LIF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001257135.1
NP_001244064.1
2
1139
NM_002309.4
NP_002300.1
3
1318
GenBank mRNA
X13967.1
-
3
1206
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel remodeling
immune response
multicellular organism development
embryo implantation
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of neuron projection development
negative regulation of angiogenesis
stem cell population maintenance
organ regeneration
positive regulation of peptidyl-serine phosphorylation
positive regulation of peptidyl-serine phosphorylation of STAT protein
tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of MAPK cascade
positive regulation of macrophage differentiation
negative regulation of meiotic nuclear division
positive regulation of transcription from RNA polymerase II promoter
decidualization
negative regulation of hormone secretion
lung alveolus development
muscle organ morphogenesis
neuron development
astrocyte differentiation
positive regulation of astrocyte differentiation
leukemia inhibitory factor signaling pathway
stem cell differentiation
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of corticotropin secretion
retina development in camera-type eye
transdifferentiation
lung vasculature development
lung lobe morphogenesis
trophoblast giant cell differentiation
spongiotrophoblast differentiation
negative regulation of ERK1 and ERK2 cascade
positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
regulation of metanephric nephron tubule epithelial cell differentiation
positive regulation of protein localization to nucleus
positive regulation of histone H3-K27 acetylation
regulation of RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor recruiting
receptor binding
cytokine activity
leukemia inhibitory factor receptor binding
protein binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0