Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNT1 SIRNA Assays ›
Gene Symbol:
WNT1
Gene Name:
Wnt family member 1
Gene Aliases:
BMND16, INT1, OI15
Chromosome Location:
Chr.12: 48978453 - 48982613 on Build GRCh38
UniGene ID:
Hs.248164
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121537
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WNT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005430.3
NP_005421.1
4
2016
Back To Top
More Information
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
embryonic axis specification
branching involved in ureteric bud morphogenesis
positive regulation of protein phosphorylation
cell-cell signaling
myoblast fusion
positive regulation of cell proliferation
response to wounding
positive regulation of lamellipodium assembly
negative regulation of cell-substrate adhesion
Wnt signaling pathway
spinal cord association neuron differentiation
diencephalon development
central nervous system morphogenesis
cerebellum formation
forebrain anterior/posterior pattern specification
midbrain-hindbrain boundary maturation during brain development
negative regulation of cell-cell adhesion
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
ubiquitin-dependent SMAD protein catabolic process
midbrain development
organ regeneration
T cell differentiation in thymus
cell proliferation in midbrain
inner ear morphogenesis
signal transduction in response to DNA damage
positive regulation of insulin-like growth factor receptor signaling pathway
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
cell fate commitment
negative regulation of fat cell differentiation
positive regulation of Notch signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fibroblast proliferation
neuron fate determination
neuron development
positive regulation of sequence-specific DNA binding transcription factor activity
Spemann organizer formation
canonical Wnt signaling pathway
Wnt signaling pathway, planar cell polarity pathway
bone development
positive regulation of dermatome development
hepatocyte differentiation
cellular response to peptide hormone stimulus
hematopoietic stem cell proliferation
positive regulation of canonical Wnt signaling pathway
negative regulation of cell aging
beta-catenin destruction complex disassembly
Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
canonical Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
embryonic brain development
negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
frizzled binding
cytokine activity
morphogen activity
protein domain specific binding
transcription regulatory region DNA binding
receptor agonist activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0