Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNFAIP3 SIRNA Assays ›
          Gene Symbol:
          TNFAIP3
          Gene Name:
          TNF alpha induced protein 3
          Gene Aliases:
          A20, AISBL, OTUD7C, TNFA1P2
          Chromosome Location:
          Chr.6: 137866317 - 137883314 on Build GRCh38
          UniGene ID:
          Hs.211600
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 121592

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNFAIP3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270507.1 NP_001257436.1 9 3384
          NM_001270508.1 NP_001257437.1 9 3408
          NM_006290.3 NP_006281.1 9 3360
          XM_005267119.1 XP_005267176.1 9 3127
          XM_006715555.1 XP_006715618.1 7 2865
          XM_011536095.1 XP_011534397.1 10 3403
          XM_011536096.1 XP_011534398.1 8 3206
          GenBank mRNA AL157444.1 - 7 2576
          BC113871.1 - 8 3061 40036694
          BC114480.1 - 8 3061 40036692
          M59465.1 - 9 3104

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          B-1 B cell homeostasis
          response to molecule of bacterial origin
          regulation of germinal center formation
          negative regulation of chronic inflammatory response
          apoptotic process
          inflammatory response
          regulation of tumor necrosis factor-mediated signaling pathway
          protein deubiquitination
          negative regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of type I interferon production
          response to muramyl dipeptide
          negative regulation of interleukin-1 beta production
          negative regulation of interleukin-2 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          negative regulation of toll-like receptor 2 signaling pathway
          negative regulation of toll-like receptor 3 signaling pathway
          negative regulation of toll-like receptor 4 signaling pathway
          negative regulation of toll-like receptor 5 signaling pathway
          protein K11-linked deubiquitination
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of protein catabolic process
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of bone resorption
          negative regulation of innate immune response
          negative regulation of smooth muscle cell proliferation
          regulation of defense response to virus by host
          regulation of inflammatory response
          negative regulation of inflammatory response
          negative regulation of B cell activation
          protein oligomerization
          regulation of vascular wound healing
          cellular response to hydrogen peroxide
          nucleotide-binding oligomerization domain containing signaling pathway
          negative regulation of nucleotide-binding oligomerization domain containing 1 signaling pathway
          negative regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
          protein K63-linked deubiquitination
          protein K48-linked ubiquitination
          protein K48-linked deubiquitination
          cellular response to lipopolysaccharide
          protein deubiquitination involved in ubiquitin-dependent protein catabolic process
          tolerance induction to lipopolysaccharide
          establishment of protein localization to vacuole
          negative regulation of osteoclast proliferation
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of cellular protein catabolic process
          positive regulation of hepatocyte proliferation
          negative regulation of CD40 signaling pathway
          negative regulation of endothelial cell apoptotic process
          protease binding
          DNA binding
          ubiquitin-protein transferase activity
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          zinc ion binding
          ligase activity
          kinase binding
          ubiquitin binding
          protein self-association
          K63-linked polyubiquitin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0