Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSP90AA1 SIRNA Assays ›
          Gene Symbol:
          HSP90AA1
          Gene Name:
          heat shock protein 90 alpha family class A member 1
          Gene Aliases:
          EL52, HEL-S-65p, HSP86, HSP89A, HSP90A, HSP90N, HSPC1, HSPCA, HSPCAL1, HSPCAL4, HSPN, Hsp89, Hsp90, LAP-2, LAP2
          Chromosome Location:
          Chr.14: 102080738 - 102139749 on Build GRCh38
          UniGene ID:
          Hs.525600
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 122318

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HSP90AA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017963.2 NP_001017963.2 10 2399
          NM_005348.3 NP_005339.3 9 1878
          XM_011536718.2 XP_011535020.1 10 2396
          GenBank mRNA AF028832.1 - 5 1563
          AJ890082.1 - 10 2336
          AJ890083.1 - 9 1728
          AK056446.1 - 10 2336
          AK291115.1 - 9 1747
          AK291607.1 - 9 1747
          BC000987.2 - 6 1138
          BC007989.2 - 5 760
          BC023006.1 - 7 1406
          BC108695.1 - 8 1734
          BC121062.2 - 9 1748 40118488
          BX247955.1 - 7 1274
          GQ149083.1 - 9 1878
          GQ472230.1 - 9 1748
          X15183.1 - 9 1748

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          Hsp90 family chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          neuron migration
          skeletal muscle contraction
          mitochondrial transport
          receptor-mediated endocytosis
          response to unfolded protein
          signal transduction
          response to heat
          response to cold
          response to salt stress
          positive regulation of lamellipodium assembly
          cardiac muscle cell apoptotic process
          peptidyl-tyrosine phosphorylation
          regulation of protein ubiquitination
          positive regulation of protein import into nucleus, translocation
          Fc-gamma receptor signaling pathway involved in phagocytosis
          ERBB2 signaling pathway
          protein refolding
          regulation of protein complex assembly
          protein unfolding
          response to estrogen
          protein import into mitochondrial outer membrane
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell size
          response to antibiotic
          vascular endothelial growth factor receptor signaling pathway
          protein stabilization
          regulation of nitric-oxide synthase activity
          chaperone-mediated protein complex assembly
          positive regulation of cardiac muscle contraction
          chaperone-mediated autophagy
          regulation of cellular response to heat
          nucleotide binding
          UTP binding
          CTP binding
          mRNA binding
          protein tyrosine kinase activity
          protein binding
          ATP binding
          GTP binding
          ATPase activity
          sulfonylurea receptor binding
          protein kinase binding
          protein phosphatase binding
          MHC class II protein complex binding
          nitric-oxide synthase regulator activity
          TPR domain binding
          dATP binding
          identical protein binding
          protein homodimerization activity
          histone deacetylase binding
          ion channel binding
          poly(A) RNA binding
          GTPase binding
          Rho GDP-dissociation inhibitor binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0