Search
Search

ECSIT| Gene Symbol: ECSIT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142464.2 | NP_001135936.1 | 4 | 678 | ||
| NM_001243204.1 | NP_001230133.1 | 4 | 688 | |||
| NM_016581.4 | NP_057665.2 | 4 | 678 | |||
| GenBank mRNA | AF243044.1 | - | 3 | 577 | ||
| AK314905.1 | - | 4 | 688 | |||
| BC000193.1 | - | 4 | 626 | 3352568 | ||
| BC005119.1 | - | 4 | 621 | 2819610 | ||
| BC008279.1 | - | 4 | 643 | 3639801 | ||
| CR457204.1 | - | 3 | 544 | |||