Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other REV3L SIRNA Assays ›
          Gene Symbol:
          REV3L
          Gene Name:
          REV3 like, DNA directed polymerase zeta catalytic subunit
          Gene Aliases:
          POLZ, REV3
          Chromosome Location:
          Chr.6: 111299031 - 111483715 on Build GRCh38
          UniGene ID:
          Hs.232021
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 12291

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: REV3L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286431.1 NP_001273360.1 6 1055
          NM_001286432.1 NP_001273361.1 5 948
          NM_002912.4 NP_002903.3 4 820
          XM_011536028.1 XP_011534330.1 3 911
          XM_011536029.2 XP_011534331.1 3 911
          XM_011536030.2 XP_011534332.1 3 911
          XM_011536031.2 XP_011534333.1 4 1039
          XM_011536032.2 XP_011534334.1 4 5629
          XM_011536036.2 XP_011534338.1 3 911
          XM_017011152.1 XP_016866641.1 4 1039
          XM_017011153.1 XP_016866642.1 4 582
          XM_017011154.1 XP_016866643.1 4 467
          XM_017011155.1 XP_016866644.1 3 911
          GenBank mRNA AF035537.1 - 2 157
          AF058701.1 - 4 447
          AF071798.1 - 3 685
          AF078695.1 - 5 950
          AF157476.1 - 3 393
          AF179428.1 - 4 521
          AF179429.1 - 5 628
          AK310949.1 - 5 944
          BC035823.1 - 3 619

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001083966, XM_006256524, XM_006256527, XM_008772988, XM_006256525, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011264, AB031049, BC048946, AF083464, AK032532.

          Panther Classification:

          Molecular Function -

          DNA metabolism protein DNA-directed DNA polymerase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA-dependent DNA replication
          error-prone translesion synthesis
          nucleic acid phosphodiester bond hydrolysis
          nucleotide binding
          DNA binding
          DNA-directed DNA polymerase activity
          protein binding
          3'-5' exonuclease activity
          metal ion binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0