Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGGY SIRNA Assays ›
Gene Symbol:
FGGY
Gene Name:
FGGY carbohydrate kinase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.1: 59296405 - 59762730 on Build GRCh38
UniGene ID:
Hs.444301
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123105
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGGY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113411.1
NP_001106882.1
7
986
NM_001244714.1
NP_001231643.1
5
623
NM_018291.3
NP_060761.3
7
986
NR_103473.1
8
944
XM_011541730.1
XP_011540032.1
7
910
XM_011541731.1
XP_011540033.1
7
844
XM_011541732.1
XP_011540034.1
7
887
XM_011541733.1
XP_011540035.1
7
805
XM_011541735.1
XP_011540037.1
7
805
XM_011541736.2
XP_011540038.1
5
751
XM_017001643.1
XP_016857132.1
7
1016
XM_017001644.1
XP_016857133.1
7
809
XM_017001645.1
XP_016857134.1
7
910
XM_017001646.1
XP_016857135.1
7
887
XM_017001647.1
XP_016857136.1
7
1015
XM_017001648.1
XP_016857137.1
8
829
XM_017001649.1
XP_016857138.1
7
955
XM_017001650.1
XP_016857139.1
9
1178
XM_017001651.1
XP_016857140.1
9
1034
XM_017001652.1
XP_016857141.1
7
955
XM_017001653.1
XP_016857142.1
7
887
XM_017001654.1
XP_016857143.1
5
706
XM_017001655.1
XP_016857144.1
5
615
XM_017001656.1
XP_016857145.1
8
1022
XM_017001657.1
XP_016857146.1
7
810
XM_017001658.1
XP_016857147.1
8
953
XM_017001659.1
XP_016857148.1
6
843
XM_017001660.1
XP_016857149.1
6
746
XM_017001661.1
XP_016857150.1
6
903
XM_017001662.1
XP_016857151.1
6
588
XM_017001663.1
XP_016857152.1
6
763
XM_017001664.1
XP_016857153.1
5
640
XM_017001665.1
XP_016857154.1
5
547
XM_017001666.1
XP_016857155.1
5
701
XM_017001667.1
XP_016857156.1
5
755
XM_017001668.1
XP_016857157.1
5
511
XM_017001669.1
XP_016857158.1
5
606
XM_017001670.1
XP_016857159.1
5
1379
XM_017001671.1
XP_016857160.1
5
507
XM_017001672.1
XP_016857161.1
6
843
XM_017001673.1
XP_016857162.1
6
900
XM_017001674.1
XP_016857163.1
6
759
XM_017001675.1
XP_016857164.1
6
843
XM_017001676.1
XP_016857165.1
6
903
XM_017001677.1
XP_016857166.1
3
3081
XM_017001678.1
XP_016857167.1
3
4356
XM_017001679.1
XP_016857168.1
7
955
GenBank mRNA
AK022237.1
-
6
843
AK054842.1
-
5
614
AK090568.1
-
8
944
AK315649.1
-
7
871
BC014947.1
-
7
871
4871664
BX647626.1
-
7
958
Back To Top
More Information
Panther Classification:
Molecular Function -
carbohydrate kinase
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
phosphorylation
neuron cellular homeostasis
molecular_function
kinase activity
phosphotransferase activity, alcohol group as acceptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-wnmjk:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline