Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGGY SIRNA Assays ›
Gene Symbol:
FGGY
Gene Name:
FGGY carbohydrate kinase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.1: 59296405 - 59762730 on Build GRCh38
UniGene ID:
Hs.444301
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123106
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGGY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113411.1
NP_001106882.1
14
1628
NM_001244714.1
NP_001231643.1
11
1193
NM_001278224.1
NP_001265153.1
7
712
NM_018291.3
NP_060761.3
13
1556
NR_103473.1
14
1514
XM_011541730.1
XP_011540032.1
14
1552
XM_011541731.1
XP_011540033.1
14
1486
XM_011541732.1
XP_011540034.1
14
1529
XM_011541733.1
XP_011540035.1
14
1447
XM_011541735.1
XP_011540037.1
13
1375
XM_011541736.2
XP_011540038.1
12
1393
XM_017001643.1
XP_016857132.1
14
1658
XM_017001644.1
XP_016857133.1
14
1451
XM_017001645.1
XP_016857134.1
13
1480
XM_017001646.1
XP_016857135.1
13
1457
XM_017001647.1
XP_016857136.1
13
1585
XM_017001648.1
XP_016857137.1
15
1471
XM_017001649.1
XP_016857138.1
14
1597
XM_017001650.1
XP_016857139.1
15
1748
XM_017001651.1
XP_016857140.1
15
1604
XM_017001652.1
XP_016857141.1
13
1525
XM_017001653.1
XP_016857142.1
13
1457
XM_017001654.1
XP_016857143.1
12
1348
XM_017001655.1
XP_016857144.1
12
1257
XM_017001656.1
XP_016857145.1
15
1664
XM_017001657.1
XP_016857146.1
14
1452
XM_017001658.1
XP_016857147.1
15
1595
XM_017001659.1
XP_016857148.1
13
1485
XM_017001660.1
XP_016857149.1
13
1388
XM_017001661.1
XP_016857150.1
13
1545
XM_017001662.1
XP_016857151.1
13
1230
XM_017001663.1
XP_016857152.1
13
1405
XM_017001664.1
XP_016857153.1
12
1282
XM_017001665.1
XP_016857154.1
12
1189
XM_017001666.1
XP_016857155.1
11
1271
XM_017001667.1
XP_016857156.1
11
1325
XM_017001668.1
XP_016857157.1
12
1153
XM_017001669.1
XP_016857158.1
12
1248
XM_017001670.1
XP_016857159.1
12
2021
XM_017001671.1
XP_016857160.1
12
1149
XM_017001672.1
XP_016857161.1
12
1413
XM_017001673.1
XP_016857162.1
12
1470
XM_017001674.1
XP_016857163.1
12
1329
XM_017001675.1
XP_016857164.1
13
1485
XM_017001676.1
XP_016857165.1
12
1473
XM_017001677.1
XP_016857166.1
10
3723
XM_017001678.1
XP_016857167.1
10
4998
GenBank mRNA
AK001848.1
-
7
586
AK054842.1
-
11
1184
AK090568.1
-
14
1514
AK315649.1
-
13
1441
BC000610.2
-
7
687
BC014947.1
-
13
1441
4871664
BX647626.1
-
14
1600
Back To Top
More Information
Panther Classification:
Molecular Function -
carbohydrate kinase
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
phosphorylation
neuron cellular homeostasis
molecular_function
kinase activity
phosphotransferase activity, alcohol group as acceptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-8xmtf:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline