Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEKHA5 SIRNA Assays ›
Gene Symbol:
PLEKHA5
Gene Name:
pleckstrin homology domain containing A5
Gene Aliases:
PEPP-2, PEPP2
Chromosome Location:
Chr.12: 19129680 - 19376400 on Build GRCh38
UniGene ID:
Hs.188614
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123166
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEKHA5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143821.2
NP_001137293.2
6
607
NM_001256470.1
NP_001243399.1
6
607
NM_001256787.1
NP_001243716.1
4
391
NM_019012.5
NP_061885.2
6
607
XM_005253400.1
XP_005253457.1
6
567
XM_006719091.1
XP_006719154.1
6
567
XM_006719093.1
XP_006719156.1
6
567
XM_006719096.1
XP_006719159.1
6
567
XM_011520714.1
XP_011519016.1
6
567
XM_011520715.1
XP_011519017.1
6
567
XM_011520716.1
XP_011519018.1
6
567
XM_011520717.1
XP_011519019.1
6
567
XM_017019495.1
XP_016874984.1
6
567
XM_017019496.1
XP_016874985.1
6
567
XM_017019497.1
XP_016874986.1
4
381
XM_017019498.1
XP_016874987.1
2
184
XM_017019499.1
XP_016874988.1
6
567
XM_017019500.1
XP_016874989.1
6
567
XM_017019501.1
XP_016874990.1
6
567
GenBank mRNA
AB051473.2
-
6
503
AB642244.1
-
6
573
AF302150.1
-
6
505
AJ270694.1
-
1
69
AK294739.1
-
4
392
AK295151.1
-
6
531
AK296277.1
-
4
384
AK297391.1
-
4
248
BC000969.2
-
1
26
BC070174.1
-
6
550
BC127091.1
-
6
505
40123820
BC127092.1
-
6
505
40123822
DC311342.1
-
4
352
DR000821.1
-
6
592
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
reproductive system development
protein binding
1-phosphatidylinositol binding
phosphatidylinositol-5-phosphate binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol-4-phosphate binding
phosphatidylinositol-3,5-bisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0