Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAQR6 SIRNA Assays ›
Gene Symbol:
PAQR6
Gene Name:
progestin and adipoQ receptor family member 6
Gene Aliases:
PRdelta
Chromosome Location:
Chr.1: 156243321 - 156248488 on Build GRCh38
UniGene ID:
Hs.235873
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123522
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAQR6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001272104.1
NP_001259033.1
8
1732
NM_001272105.1
NP_001259034.1
9
1804
NM_001272106.1
NP_001259035.1
6
1698
NM_001272107.1
NP_001259036.1
7
1867
NM_001272108.1
NP_001259037.1
6
1357
NM_001272109.1
NP_001259038.1
5
1622
NM_001272110.1
NP_001259039.1
7
1605
NM_001272111.1
NP_001259040.1
7
1563
NM_001272112.1
NP_001259041.1
5
1390
NM_001272113.1
NP_001259042.1
5
1260
NM_024897.3
NP_079173.2
7
1938
NM_198406.2
NP_940798.1
8
1690
NR_073610.1
7
1614
XM_005245494.3
XP_005245551.1
6
1858
XM_006711546.2
XP_006711609.1
8
1701
XM_006711547.3
XP_006711610.1
8
2174
XM_006711548.3
XP_006711611.1
7
2056
XM_006711552.2
XP_006711615.1
6
1799
XM_006711553.3
XP_006711616.1
6
1841
XM_011510000.2
XP_011508302.1
7
1964
XM_017002385.1
XP_016857874.1
7
3332
GenBank mRNA
AA310453.1
-
1
46
AF455045.1
-
7
1785
AF455046.1
-
5
1567
AF455047.1
-
7
1529
AK026325.1
-
4
1130
AK054682.1
-
7
1544
AK094346.1
-
7
1583
AK095143.1
-
8
1662
AK095197.1
-
7
1873
AK307441.1
-
6
1394
AL834227.1
-
7
1758
AY424284.1
-
8
1611
BC058509.1
-
8
1574
5770270
BC071623.1
-
3
851
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
steroid hormone mediated signaling pathway
response to steroid hormone
steroid hormone receptor activity
steroid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0