Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAP1GAP SIRNA Assays ›
          Gene Symbol:
          RAP1GAP
          Gene Name:
          RAP1 GTPase activating protein
          Gene Aliases:
          RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
          Chromosome Location:
          Chr.1: 21596215 - 21669444 on Build GRCh38
          UniGene ID:
          Hs.148178
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 12364

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAP1GAP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145657.1 NP_001139129.1 8, 9 664
          NM_001145658.1 NP_001139130.1 7, 8 524
          NM_002885.2 NP_002876.2 7, 8 557
          XM_005245955.3 XP_005246012.2 7, 8 569
          XM_006710804.2 XP_006710867.2 7, 8 569
          XM_006710805.2 XP_006710868.2 7, 8 569
          XM_017001965.1 XP_016857454.1 6, 7 571
          XM_017001966.1 XP_016857455.1 6, 7 571
          XM_017001967.1 XP_016857456.1 7, 8 569
          XM_017001968.1 XP_016857457.1 7, 8 604
          XM_017001969.1 XP_016857458.1 6, 7 571
          XM_017001970.1 XP_016857459.1 6, 7 571
          XM_017001971.1 XP_016857460.1 7, 8 523
          XM_017001972.1 XP_016857461.1 7, 8 569
          XM_017001973.1 XP_016857462.1 6, 7 602
          XM_017001974.1 XP_016857463.1 7, 8 569
          XM_017001975.1 XP_016857464.1 6, 7 571
          XM_017001976.1 XP_016857465.1 7, 8 569
          XM_017001977.1 XP_016857466.1 7, 8 569
          XM_017001978.1 XP_016857467.1 8, 9 718
          XM_017001979.1 XP_016857468.1 9, 10 722
          XM_017001980.1 XP_016857469.1 7, 8 523
          XM_017001981.1 XP_016857470.1 9, 10 698
          XM_017001982.1 XP_016857471.1 8, 9 718
          XM_017001983.1 XP_016857472.1 10, 11 930
          XM_017001984.1 XP_016857473.1 9, 10 698
          XM_017001985.1 XP_016857474.1 7, 8 523
          XM_017001986.1 XP_016857475.1 9, 10 735
          XM_017001987.1 XP_016857476.1 8, 9 664
          XM_017001988.1 XP_016857477.1 8, 9 635
          XM_017001989.1 XP_016857478.1 9, 10 735
          XM_017001990.1 XP_016857479.1 8, 9 620
          XM_017001991.1 XP_016857480.1 8, 9 658
          GenBank mRNA AB007943.3 - 8, 9 602
          BC035030.1 - 4, 5 313 5261027
          BC054490.1 - 7, 8 464 5767775
          BP200029.1 - 7, 8 524
          BP283134.1 - 7, 8 557
          CV574545.1 - 7, 8 514
          M64788.1 - 7, 8 500

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          regulation of GTPase activity
          positive regulation of GTPase activity
          negative regulation of neuron differentiation
          regulation of small GTPase mediated signal transduction
          negative regulation of microvillus assembly
          negative regulation of GTP binding
          negative regulation of thyroid gland epithelial cell proliferation
          cellular response to glial cell derived neurotrophic factor
          GTPase activity
          GTPase activator activity
          protein binding
          Ras GTPase binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0