Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG2 SIRNA Assays ›
Gene Symbol:
DLG2
Gene Name:
discs large MAGUK scaffold protein 2
Gene Aliases:
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location:
Chr.11: 83455009 - 85628534 on Build GRCh38
UniGene ID:
Hs.367656
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124292
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142699.1
NP_001136171.1
28
6321
NM_001142700.1
NP_001136172.1
20
5356
NM_001142702.1
NP_001136174.1
11
4398
NM_001206769.1
NP_001193698.1
22
6005
NM_001300983.1
NP_001287912.1
23
5943
NM_001364.3
NP_001355.2
23
5997
XM_005273810.4
XP_005273867.1
22
5640
XM_005273811.4
XP_005273868.1
22
5586
XM_011544778.2
XP_011543080.1
29
7612
XM_011544780.2
XP_011543082.1
29
7558
XM_011544782.2
XP_011543084.1
24
6083
XM_011544788.2
XP_011543090.1
21
5430
XM_017017254.1
XP_016872743.1
30
7654
XM_017017255.1
XP_016872744.1
29
7615
XM_017017256.1
XP_016872745.1
29
7144
XM_017017257.1
XP_016872746.1
28
8076
XM_017017258.1
XP_016872747.1
27
7252
XM_017017259.1
XP_016872748.1
27
6973
XM_017017260.1
XP_016872749.1
26
7905
XM_017017261.1
XP_016872750.1
28
7402
XM_017017262.1
XP_016872751.1
23
6107
XM_017017263.1
XP_016872752.1
25
6125
XM_017017264.1
XP_016872753.1
24
6302
XM_017017265.1
XP_016872754.1
24
6299
XM_017017266.1
XP_016872755.1
22
6009
XM_017017267.1
XP_016872756.1
24
6207
XM_017017268.1
XP_016872757.1
24
6028
XM_017017269.1
XP_016872758.1
24
6729
XM_017017270.1
XP_016872759.1
24
6029
XM_017017271.1
XP_016872760.1
23
6206
XM_017017272.1
XP_016872761.1
23
6203
XM_017017273.1
XP_016872762.1
22
5958
XM_017017274.1
XP_016872763.1
24
6153
XM_017017275.1
XP_016872764.1
21
5908
XM_017017276.1
XP_016872765.1
23
6631
XM_017017277.1
XP_016872766.1
23
5951
XM_017017278.1
XP_016872767.1
21
5855
XM_017017279.1
XP_016872768.1
23
5873
XM_017017280.1
XP_016872769.1
23
6550
XM_017017281.1
XP_016872770.1
22
6050
XM_017017283.1
XP_016872772.1
22
5855
XM_017017284.1
XP_016872773.1
22
5527
XM_017017285.1
XP_016872774.1
22
6477
XM_017017286.1
XP_016872775.1
20
5391
XM_017017287.1
XP_016872776.1
13
4411
XM_017017288.1
XP_016872777.1
12
4250
XM_017017289.1
XP_016872778.1
12
5825
XM_017017290.1
XP_016872779.1
12
4470
XM_017017291.1
XP_016872780.1
11
4217
XM_017017292.1
XP_016872781.1
11
5729
GenBank mRNA
AK094585.1
-
1
419
AK125151.1
-
1
1427
AK125152.1
-
1
950
BC104969.1
-
1
518
BC110844.1
-
1
773
BX537627.1
-
1
308
BX648386.1
-
1
1824
CR933674.1
-
11
4367
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
nervous system development
negative regulation of phosphatase activity
sensory perception of pain
receptor clustering
establishment or maintenance of epithelial cell apical/basal polarity
GMP metabolic process
GDP metabolic process
receptor localization to synapse
guanylate kinase activity
protein binding
kinase binding
ionotropic glutamate receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0