Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASCC2 SIRNA Assays ›
Gene Symbol:
ASCC2
Gene Name:
activating signal cointegrator 1 complex subunit 2
Gene Aliases:
ASC1p100, p100
Chromosome Location:
Chr.22: 29788608 - 29838304 on Build GRCh38
UniGene ID:
Hs.731754
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125680
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASCC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242906.1
NP_001229835.1
5
485
NM_032204.4
NP_115580.2
6
713
XM_005261775.2
XP_005261832.1
7
753
XM_011530442.2
XP_011528744.1
7
753
XM_011530443.2
XP_011528745.1
8
819
XM_011530444.2
XP_011528746.1
8
866
XM_011530445.2
XP_011528747.1
7
800
XM_011530446.2
XP_011528748.1
6
683
XM_011530448.2
XP_011528750.1
6
586
XM_011530449.2
XP_011528751.1
5
524
XM_011530450.2
XP_011528752.1
8
869
XM_011530451.2
XP_011528753.1
9
943
XM_011530452.2
XP_011528754.1
8
916
XM_011530453.2
XP_011528755.1
7
771
XM_011530454.2
XP_011528756.1
6
655
XM_011530455.2
XP_011528757.1
5
584
XM_017028991.1
XP_016884480.1
6
2891
XM_017028992.1
XP_016884481.1
6
2891
XM_017028993.1
XP_016884482.1
6
2891
XM_017028994.1
XP_016884483.1
6
2891
XM_017028995.1
XP_016884484.1
5
2732
XM_017028996.1
XP_016884485.1
7
1693
XM_017028997.1
XP_016884486.1
5
2732
XM_017028998.1
XP_016884487.1
7
1693
XM_017028999.1
XP_016884488.1
8
819
XM_017029000.1
XP_016884489.1
7
800
XM_017029001.1
XP_016884490.1
6
586
XM_017029002.1
XP_016884491.1
5
524
XM_017029003.1
XP_016884492.1
7
660
XM_017029004.1
XP_016884493.1
5
524
XM_017029005.1
XP_016884494.1
8
869
XM_017029006.1
XP_016884495.1
9
943
XM_017029007.1
XP_016884496.1
7
771
XM_017029008.1
XP_016884497.1
6
655
XM_017029009.1
XP_016884498.1
5
584
GenBank mRNA
AK022886.1
-
8
831
AK303257.1
-
5
455
AY013289.1
-
5
590
BC025368.2
-
6
674
3955747
BP201867.1
-
5
560
Back To Top
More Information
Gene Ontology Categories:
Function(s)
DNA dealkylation involved in DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0