Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCL5 SIRNA Assays ›
          Gene Symbol:
          CCL5
          Gene Name:
          C-C motif chemokine ligand 5
          Gene Aliases:
          D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP
          Chromosome Location:
          Chr.17: 35871491 - 35880373 on Build GRCh38
          UniGene ID:
          Hs.514821
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 12607

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCL5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278736.1 NP_001265665.1 4 955
          NM_002985.2 NP_002976.2 3 873
          GenBank mRNA AA486131.1 - 1 165
          M21121.1 - 3 833

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          dendritic cell chemotaxis
          chronic inflammatory response
          monocyte chemotaxis
          regulation of chronic inflammatory response
          calcium ion transport
          cellular calcium ion homeostasis
          exocytosis
          chemotaxis
          inflammatory response
          leukocyte cell-cell adhesion
          G-protein coupled receptor signaling pathway
          cell-cell signaling
          aging
          response to virus
          response to toxic substance
          response to salt stress
          positive regulation of activation of JAK2 kinase activity
          positive regulation of macrophage chemotaxis
          positive regulation of T cell chemotaxis
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to activity
          positive regulation of smooth muscle cell migration
          dibenzo-p-dioxin metabolic process
          positive regulation of cell migration
          neutrophil chemotaxis
          positive regulation of cellular biosynthetic process
          activation of phospholipase D activity
          positive regulation of fever generation
          lipopolysaccharide-mediated signaling pathway
          response to insulin
          positive regulation of cell-cell adhesion mediated by integrin
          positive regulation of homotypic cell-cell adhesion
          positive regulation of T cell proliferation
          neutrophil activation
          positive regulation of phosphorylation
          response to drug
          positive regulation of tyrosine phosphorylation of STAT protein
          protein kinase B signaling
          positive regulation of GTPase activity
          cellular protein complex assembly
          response to estrogen
          negative regulation by host of viral transcription
          cellular response to fibroblast growth factor stimulus
          positive regulation of viral genome replication
          negative regulation of viral genome replication
          positive regulation of innate immune response
          positive regulation of neuron differentiation
          positive regulation of osteoclast differentiation
          negative regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          positive regulation of translational initiation
          positive regulation of JAK-STAT cascade
          eosinophil chemotaxis
          macrophage chemotaxis
          lymphocyte chemotaxis
          positive regulation of smooth muscle cell proliferation
          positive regulation of epithelial cell proliferation
          positive regulation of inflammatory response
          regulation of insulin secretion
          regulation of T cell activation
          positive chemotaxis
          protein tetramerization
          response to glucocorticoid
          positive regulation of calcium ion transport
          positive regulation of mast cell chemotaxis
          positive regulation of protein tyrosine kinase activity
          chemokine-mediated signaling pathway
          negative regulation of chemokine-mediated signaling pathway
          negative regulation of T cell apoptotic process
          positive regulation of T cell apoptotic process
          positive regulation of ERK1 and ERK2 cascade
          response to cholesterol
          cellular response to amino acid stimulus
          cellular response to vitamin K
          cellular response to morphine
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to ethanol
          cellular response to high density lipoprotein particle stimulus
          cellular response to organic cyclic compound
          cellular response to alkyl hydroperoxide
          cellular response to transforming growth factor beta stimulus
          positive regulation of monocyte chemotaxis
          regulation of neuron death
          negative regulation of neuron death
          negative regulation of macrophage apoptotic process
          positive regulation of T cell migration
          positive regulation of natural killer cell chemotaxis
          phosphatidylinositol phospholipase C activity
          protein kinase activity
          protein binding
          chemokine activity
          heparin binding
          phospholipase activator activity
          receptor signaling protein tyrosine kinase activator activity
          CCR1 chemokine receptor binding
          CCR4 chemokine receptor binding
          CCR5 chemokine receptor binding
          chemoattractant activity
          chemokine receptor binding
          protein homodimerization activity
          protein self-association
          chemokine receptor antagonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0