Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPIP5K2 SIRNA Assays ›
          Gene Symbol:
          PPIP5K2
          Gene Name:
          diphosphoinositol pentakisphosphate kinase 2
          Gene Aliases:
          CFAP160, HISPPD1, IP7K2, VIP2
          Chromosome Location:
          Chr.5: 103120254 - 103203523 on Build GRCh38
          UniGene ID:
          Hs.212046
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 126807

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPIP5K2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276277.1 NP_001263206.1 3 710
          NM_001281471.1 NP_001268400.1 3 710
          NM_015216.3 NP_056031.2 2 475
          XM_005271935.1 XP_005271992.1 3 709
          XM_005271936.1 XP_005271993.1 3 709
          XM_005271938.1 XP_005271995.1 3 709
          XM_005277534.1 XP_005277591.1 3 709
          XM_005277535.1 XP_005277592.1 3 709
          XM_005277536.1 XP_005277593.1 3 709
          XM_005277538.1 XP_005277595.1 3 709
          XM_005277539.1 XP_005277596.1 3 709
          XM_005277541.1 XP_005277598.1 3 709
          XM_006714576.1 XP_006714639.1 3 709
          XM_006714577.1 XP_006714640.1 3 709
          XM_011543290.2 XP_011541592.1 3 709
          XM_017009269.1 XP_016864758.1 3 709
          XM_017009270.1 XP_016864759.1 3 709
          XM_017009271.1 XP_016864760.1 3 709
          XM_017009272.1 XP_016864761.1 3 709
          XM_017009273.1 XP_016864762.1 3 709
          XM_017009274.1 XP_016864763.1 3 709
          XM_017009275.1 XP_016864764.1 3 709
          GenBank mRNA AB007893.1 - 3 700
          AK294787.1 - 3 574
          BC024591.1 - 2 228 3913521

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          inositol metabolic process
          phosphorylation
          dephosphorylation
          inositol phosphate biosynthetic process
          inositol phosphate metabolic process
          inositol-1,3,4,5,6-pentakisphosphate kinase activity
          inositol hexakisphosphate kinase activity
          inositol heptakisphosphate kinase activity
          inositol hexakisphosphate 5-kinase activity
          acid phosphatase activity
          ATP binding
          diphosphoinositol-pentakisphosphate kinase activity
          inositol hexakisphosphate 1-kinase activity
          inositol hexakisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0