Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTR3B SIRNA Assays ›
          Gene Symbol:
          ACTR3B ACTR3C
          Gene Name:
          ARP3 actin related protein 3 homolog B
          ARP3 actin-related protein 3 homolog C
          Gene Aliases:
          ARP11, ARP3BETA
          ARP11
          Chromosome Location:
          Chr.7: 152759749 - 153070979 on Build GRCh38
          Chromosome Location:
          Chr.7: 149881359 - 150325523 on Build GRCh38
          UniGene ID:
          Hs.647117
          Hs.664579
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 127243

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACTR3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040135.2 NP_001035225.1 7 731
          NM_020445.5 NP_065178.1 7 731
          NR_073000.1 6 675
          NR_073001.1 6 606
          XM_006716065.2 XP_006716128.1 7 713
          XM_006716066.1 XP_006716129.1 7 713
          XM_006716067.3 XP_006716130.1 7 713
          XM_011516425.2 XP_011514727.1 7 713
          XM_011516426.2 XP_011514728.1 7 713
          XM_011516427.1 XP_011514729.1 7 713
          XM_017012460.1 XP_016867949.1 7 713
          XM_017012461.1 XP_016867950.1 7 713
          XM_017012462.1 XP_016867951.1 7 642
          XM_017012463.1 XP_016867952.1 7 581
          XM_017012464.1 XP_016867953.1 6 672
          XM_017012465.1 XP_016867954.1 6 535
          GenBank mRNA AB209174.1 - 3 4058
          AF023453.1 - 7 633
          AF086920.1 - 6 597
          AK001198.1 - 3 199
          AK294292.1 - 6 629
          AK315534.1 - 7 713
          AK316078.1 - 6 526
          BC008682.1 - 7 686 3854008
          BC015207.1 - 7 643 3916310
          DN601600.1 - 7 731
          Gene Symbol: ACTR3C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164458.1 NP_001157930.1 4 462
          NM_001164459.1 NP_001157931.1 4 400
          XM_005250043.4 XP_005250100.3 4 462
          XM_011516502.2 XP_011514804.1 4 462
          XM_011516506.2 XP_011514808.1 4 462
          XM_011516507.2 XP_011514809.1 4 462
          XM_011516508.2 XP_011514810.1 4 462
          XM_011516511.2 XP_011514813.1 4 462
          XM_017012548.1 XP_016868037.1 4 462
          XM_017012549.1 XP_016868038.1 4 462
          XM_017012550.1 XP_016868039.1 4 462
          XM_017012551.1 XP_016868040.1 4 462
          XM_017012553.1 XP_016868042.1 4 2312
          GenBank mRNA AB039791.1 - 4 400
          BX640643.1 - 3 204
          DA868104.1 - 4 462
          DN992136.1 - 4 526
          DR004860.1 - 4 303

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001191860.
          This siRNA also targets the following transcripts in Mouse: NM_001004365, BC053106, BC082279, AK035798.

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          Arp2/3 complex-mediated actin nucleation
          molecular_function
          actin binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0