Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CFAP69 SIRNA Assays ›
Gene Symbol:
CFAP69
Gene Name:
cilia and flagella associated protein 69
Gene Aliases:
C7orf63, FAP69
Chromosome Location:
Chr.7: 90245174 - 90320964 on Build GRCh38
UniGene ID:
Hs.521014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
127457
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CFAP69
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039706.2
NP_001034795.2
14, 15
1915
NM_001160138.1
NP_001153610.1
14, 15
1861
XM_005250600.3
XP_005250657.1
15, 16
1920
XM_011516577.2
XP_011514879.1
14, 15
1894
XM_011516578.2
XP_011514880.1
14, 15
1894
XM_017012624.1
XP_016868113.1
14, 15
1840
XM_017012625.1
XP_016868114.1
14, 15
1894
XM_017012626.1
XP_016868115.1
14, 15
1894
XM_017012627.1
XP_016868116.1
14, 15
1894
XM_017012628.1
XP_016868117.1
14, 15
1894
XM_017012629.1
XP_016868118.1
13, 14
1784
XM_017012630.1
XP_016868119.1
14, 15
1894
XM_017012631.1
XP_016868120.1
12, 13
1442
XM_017012632.1
XP_016868121.1
14, 15
1894
XM_017012633.1
XP_016868122.1
12, 13
4471
XM_017012634.1
XP_016868123.1
12, 13
1609
XM_017012635.1
XP_016868124.1
10, 11
1305
XM_017012636.1
XP_016868125.1
9, 10
1246
XM_017012637.1
XP_016868126.1
10, 11
4411
XM_017012638.1
XP_016868127.1
10, 11
1252
XM_017012639.1
XP_016868128.1
10, 11
1287
XM_017012640.1
XP_016868129.1
7, 8
891
XM_017012641.1
XP_016868130.1
14, 15
1894
GenBank mRNA
AK024715.1
-
3, 4
413
AK093991.1
-
12, 13
1697
AK302636.1
-
14, 15
1820
AK303034.1
-
14, 15
1894
AL833446.1
-
8, 9
1108
BC036351.1
-
15, 16
2079
BC132771.1
-
8, 9
1137
BC140742.1
-
14, 15
1801
9021610
BC141834.1
-
14, 15
1861
40148850
BC142655.1
-
14, 15
1861
40148060
Back To Top
More Information
Panther Classification:
Molecular Function -
structural protein
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-glv4n:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0