Search
Search

DPY19L4| Gene Symbol: DPY19L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181787.2 | NP_861452.2 | 9 | 1108 | ||
| XM_005250894.4 | XP_005250951.2 | 9 | 1321 | |||
| XM_005250895.3 | XP_005250952.1 | 8 | 989 | |||
| XM_011516983.2 | XP_011515285.1 | 9 | 1114 | |||
| XM_011516984.2 | XP_011515286.1 | 8 | 988 | |||
| XM_017013352.1 | XP_016868841.1 | 7 | 1059 | |||
| XM_017013354.1 | XP_016868843.1 | 8 | 1014 | |||
| XM_017013355.1 | XP_016868844.1 | 8 | 1007 | |||
| XM_017013356.1 | XP_016868845.1 | 7 | 860 | |||
| GenBank mRNA | AK123618.1 | - | 1 | 103 | ||
| AK123682.1 | - | 9 | 1062 | |||
| AL832859.1 | - | 4 | 423 | |||
| BC110870.1 | - | 9 | 1036 | 5580071 | ||
| BC126193.1 | - | 9 | 1004 | 8991909 | ||
| BC130576.1 | - | 9 | 1004 | 40146607 | ||
| BX538048.1 | - | 9 | 1056 | |||
| BX538174.1 | - | 9 | 1060 | |||