Search
Search

SLAMF6| Gene Symbol: SLAMF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184714.1 | NP_001171643.1 | 2 | 306 | ||
| NM_001184715.1 | NP_001171644.1 | 2 | 159 | |||
| NM_052931.4 | NP_443163.1 | 2 | 306 | |||
| XM_017000215.1 | XP_016855704.1 | 2 | 312 | |||
| XM_017000216.1 | XP_016855705.1 | 2 | 132 | |||
| GenBank mRNA | AJ277141.1 | - | 2 | 248 | ||
| AJ306388.1 | - | 2 | 245 | |||
| AK125624.1 | - | 2 | 306 | |||
| AK303990.1 | - | 2 | 152 | |||
| AK310085.1 | - | 2 | 132 | |||
| AK313549.1 | - | 2 | 258 | |||
| AL832854.1 | - | 2 | 287 | |||
| AY358159.1 | - | 2 | 297 | |||
| BC113893.1 | - | 2 | 263 | 40083595 | ||
| BC114495.1 | - | 2 | 263 | 40083594 | ||
| DB197632.1 | - | 2 | 306 | |||