Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SPDYE1 SIRNA Assays ›
          Gene Symbol:
          SPDYE1 SPDYE16 SPDYE18 SPDYE2 SPDYE2B SPDYE5 SPDYE6
          Gene Name:
          speedy/RINGO cell cycle regulator family member E1
          speedy/RINGO cell cycle regulator family member E16
          speedy/RINGO cell cycle regulator family member E18
          speedy/RINGO cell cycle regulator family member E2
          speedy/RINGO cell cycle regulator family member E2B
          speedy/RINGO cell cycle regulator family member E5
          speedy/RINGO cell cycle regulator family member E6
          Gene Aliases:
          Ringo1, SPDYE, WBSCR19
          -
          -
          SPDYB2-L1
          SPDYE2L
          -
          -
          Chromosome Location:
          Chr.7: 43997855 - 44011040 on Build GRCh38
          Chromosome Location:
          Chr.7: 76532626 - 76540660 on Build GRCh38
          Chromosome Location:
          Chr.7: 77051689 - 77060537 on Build GRCh38
          Chromosome Location:
          Chr.7: 102551232 - 102565227 on Build GRCh38
          Chromosome Location:
          Chr.7: 102650325 - 102663525 on Build GRCh38
          Chromosome Location:
          Chr.7: 75493627 - 75504304 on Build GRCh38
          Chromosome Location:
          Chr.7: 102345752 - 102356444 on Build GRCh38
          UniGene ID:
          Hs.645483
          -
          -
          Hs.656428
          Hs.433879
          Hs.632298
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 129882

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SPDYE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175064.2 NP_778234.2 1 339
          XM_005249719.3 XP_005249776.1 3 850
          XM_017012015.1 XP_016867504.1 3 788
          XM_017012016.1 XP_016867505.1 2 200
          GenBank mRNA AF412027.1 - 1 348
          AL137266.1 - 1 121
          Gene Symbol: SPDYE16
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006716211.3 XP_006716274.1 1 477
          XM_017012887.1 XP_016868376.1 1 477
          Gene Symbol: SPDYE18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005250742.4 XP_005250799.1 2 348
          XM_005250743.4 XP_005250800.1 1 203
          XM_011516756.2 XP_011515058.1 2 348
          Gene Symbol: SPDYE2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001031618.3 NP_001026789.2 3 797
          XM_011516237.2 XP_011514539.1 3 796
          XM_017012223.1 XP_016867712.1 3 796
          GenBank mRNA AK302217.1 - 3 797
          JQ023160.1 - 2 323
          Gene Symbol: SPDYE2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166339.1 NP_001159811.1 3 797
          XM_005250093.3 XP_005250150.1 2 1365
          XM_011515702.2 XP_011514004.1 2 1365
          Gene Symbol: SPDYE5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306141.1 NP_001293070.1 2 744
          Gene Symbol: SPDYE6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146210.2 NP_001139682.2 2 744

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein kinase activity
          protein kinase binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0