Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFX SIRNA Assays ›
Gene Symbol:
ZFX
Gene Name:
zinc finger protein, X-linked
Gene Aliases:
ZNF926
Chromosome Location:
Chr.X: 24148994 - 24216255 on Build GRCh38
UniGene ID:
Hs.336681
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13034
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178084.1
NP_001171555.1
3
710
NM_001178085.1
NP_001171556.1
6
1054
NM_001178095.1
NP_001171566.1
3
789
NM_003410.3
NP_003401.2
5
918
XM_005274591.4
XP_005274648.1
4
1084
XM_005274592.3
XP_005274649.1
3
899
XM_005274594.4
XP_005274651.1
2
2089
XM_006724513.3
XP_006724576.1
6
1313
XM_006724515.3
XP_006724578.1
6
1315
XM_011545578.1
XP_011543880.1
6
1195
XM_011545579.1
XP_011543881.1
5
983
XM_011545581.2
XP_011543883.1
5
1125
XM_011545583.2
XP_011543885.1
2
639
XM_017029788.1
XP_016885277.1
6
1236
XM_017029789.1
XP_016885278.1
5
1798
XM_017029790.1
XP_016885279.1
5
1203
XM_017029791.1
XP_016885280.1
4
1093
XM_017029792.1
XP_016885281.1
4
972
XM_017029793.1
XP_016885282.1
3
770
XM_017029794.1
XP_016885283.1
5
1024
XM_017029795.1
XP_016885284.1
2
653
XM_017029796.1
XP_016885285.1
4
805
XM_017029797.1
XP_016885286.1
6
1264
XM_017029798.1
XP_016885287.1
2
884
XM_017029799.1
XP_016885288.1
3
899
XM_017029800.1
XP_016885289.1
2
886
GenBank mRNA
AB209892.1
-
3
822
AF505615.1
-
2
611
AF505616.1
-
2
611
AF505618.1
-
2
611
AF505619.1
-
2
611
BC136308.1
-
5
906
9020295
BC136309.1
-
5
906
9020296
M30608.1
-
4
869
X59739.1
-
3
689
X59740.1
-
6
1038
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0