Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT2 SIRNA Assays ›
          Gene Symbol:
          WNT2
          Gene Name:
          Wnt family member 2
          Gene Aliases:
          INT1L1, IRP
          Chromosome Location:
          Chr.7: 117276631 - 117323289 on Build GRCh38
          UniGene ID:
          Hs.567356
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 13116

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003391.2 NP_003382.1 2 421
          NR_024047.1 2 421
          GenBank mRNA AK056742.1 - 2 418
          BC029854.1 - 2 173
          BC078170.1 - 2 407 5761421
          BT019608.1 - 2 121
          X07876.1 - 2 415

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          positive regulation of mesenchymal cell proliferation
          lens development in camera-type eye
          cell-cell signaling
          positive regulation of cell proliferation
          Wnt signaling pathway
          neurogenesis
          neuron differentiation
          lung development
          cell proliferation in midbrain
          cell fate commitment
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          positive regulation of neurogenesis
          positive regulation of sequence-specific DNA binding transcription factor activity
          atrial cardiac muscle tissue morphogenesis
          positive regulation of cardiac muscle cell proliferation
          canonical Wnt signaling pathway
          cardiac epithelial to mesenchymal transition
          lung induction
          positive regulation of epithelial cell proliferation involved in lung morphogenesis
          labyrinthine layer blood vessel development
          iris morphogenesis
          mammary gland epithelium development
          cellular response to retinoic acid
          cellular response to transforming growth factor beta stimulus
          positive regulation of canonical Wnt signaling pathway
          midbrain dopaminergic neuron differentiation
          canonical Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
          frizzled binding
          cytokine activity
          protein binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0