Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OBSCN SIRNA Assays ›
          Gene Symbol:
          OBSCN
          Gene Name:
          obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
          Gene Aliases:
          ARHGEF30, UNC89
          Chromosome Location:
          Chr.1: 228208013 - 228381431 on Build GRCh38
          UniGene ID:
          Hs.650039
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 131675

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OBSCN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098623.2 NP_001092093.2 3, 4 1340
          NM_001271223.2 NP_001258152.2 3, 4 1340
          NM_052843.3 NP_443075.3 3, 4 1340
          XM_005273287.4 XP_005273344.1 3, 4 1420
          XM_005273291.4 XP_005273348.1 3, 4 1420
          XM_005273298.4 XP_005273355.1 3, 4 1420
          XM_005273307.4 XP_005273364.1 3, 4 1420
          XM_006711822.3 XP_006711885.1 3, 4 1420
          XM_006711823.3 XP_006711886.1 3, 4 1420
          XM_006711827.3 XP_006711890.1 3, 4 1419
          XM_006711829.3 XP_006711892.1 3, 4 1419
          XM_011544297.2 XP_011542599.1 3, 4 1420
          XM_011544299.2 XP_011542601.1 3, 4 1420
          XM_017002443.1 XP_016857932.1 3, 4 1420
          XM_017002444.1 XP_016857933.1 3, 4 1383
          XM_017002445.1 XP_016857934.1 3, 4 1420
          XM_017002446.1 XP_016857935.1 3, 4 1420
          XM_017002447.1 XP_016857936.1 3, 4 1420
          XM_017002448.1 XP_016857937.1 3, 4 1420
          XM_017002449.1 XP_016857938.1 3, 4 1421
          XM_017002450.1 XP_016857939.1 3, 4 1421
          XM_017002451.1 XP_016857940.1 3, 4 1421
          XM_017002452.1 XP_016857941.1 3, 4 1421
          XM_017002453.1 XP_016857942.1 3, 4 1420
          XM_017002454.1 XP_016857943.1 3, 4 1420
          XM_017002455.1 XP_016857944.1 3, 4 1420
          XM_017002456.1 XP_016857945.1 3, 4 1420
          XM_017002457.1 XP_016857946.1 3, 4 1420
          XM_017002458.1 XP_016857947.1 3, 4 1421
          XM_017002459.1 XP_016857948.1 3, 4 1421
          XM_017002460.1 XP_016857949.1 3, 4 1421
          XM_017002461.1 XP_016857950.1 3, 4 1420
          XM_017002462.1 XP_016857951.1 3, 4 1420
          XM_017002463.1 XP_016857952.1 3, 4 1420
          XM_017002464.1 XP_016857953.1 3, 4 1420
          XM_017002465.1 XP_016857954.1 3, 4 1421
          XM_017002466.1 XP_016857955.1 3, 4 1421
          XM_017002467.1 XP_016857956.1 3, 4 1421
          XM_017002468.1 XP_016857957.1 3, 4 1421
          XM_017002469.1 XP_016857958.1 3, 4 1421
          XM_017002470.1 XP_016857959.1 3, 4 1422
          XM_017002471.1 XP_016857960.1 3, 4 1419
          XM_017002472.1 XP_016857961.1 3, 4 1419
          XM_017002473.1 XP_016857962.1 3, 4 1419
          GenBank mRNA AJ002535.1 - 3, 4 1337

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          multicellular organism development
          regulation of Rho protein signal transduction
          protein localization to M-band
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          sarcomere organization
          regulation of small GTPase mediated signal transduction
          mitophagy in response to mitochondrial depolarization
          protein serine/threonine kinase activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          calmodulin binding
          ATP binding
          structural constituent of muscle
          ankyrin binding
          titin binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline