Search
Search

IGSF8| Gene Symbol: IGSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206665.2 | NP_001193594.1 | 6 | 1660 | ||
| NM_001320247.1 | NP_001307176.1 | 5 | 1802 | |||
| NM_052868.5 | NP_443100.1 | 5 | 1802 | |||
| XM_006711631.3 | XP_006711694.1 | 5 | 1551 | |||
| XM_011510159.2 | XP_011508461.1 | 5 | 2182 | |||
| XM_017002835.1 | XP_016858324.1 | 6 | 1666 | |||
| XM_017002836.1 | XP_016858325.1 | 6 | 1857 | |||
| XM_017002837.1 | XP_016858326.1 | 6 | 1666 | |||
| XM_017002838.1 | XP_016858327.1 | 5 | 3802 | |||
| XM_017002839.1 | XP_016858328.1 | 6 | 3714 | |||
| XM_017002840.1 | XP_016858329.1 | 6 | 1828 | |||
| GenBank mRNA | AF407274.1 | - | 5 | 1742 | ||
| AK055843.1 | - | 5 | 1538 | |||
| AK090396.1 | - | 1 | 4594 | |||
| AK308115.1 | - | 6 | 1661 | |||
| AY044845.1 | - | 5 | 1657 | |||
| AY157579.1 | - | 5 | 1611 | |||
| BC004108.2 | - | 4 | 1199 | |||
| BC011896.2 | - | 2 | 680 | |||
| BC023649.2 | - | 3 | 867 | |||
| BC053881.1 | - | 5 | 1748 | 5752209 | ||