Search
Search

EFCAB11| Gene Symbol: EFCAB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284266.1 | NP_001271195.1 | 4 | 386 | ||
| NM_001284267.1 | NP_001271196.1 | 4 | 239 | |||
| NM_001284268.1 | NP_001271197.1 | 4 | 397 | |||
| NM_001284269.1 | NP_001271198.1 | 4 | 397 | |||
| NM_145231.3 | NP_660274.1 | 4 | 386 | |||
| GenBank mRNA | AK094740.1 | - | 4 | 339 | ||
| AK298930.1 | - | 4 | 331 | |||
| AK310331.1 | - | 3 | 298 | |||
| AL832321.1 | - | 4 | 323 | |||
| BC001787.1 | - | 4 | 298 | 3353520 | ||
| BG494945.1 | - | 4 | 351 | |||
| BP371056.1 | - | 4 | 239 | |||
| BX161414.1 | - | 4 | 363 | |||
| BX247991.1 | - | 4 | 304 | |||
| CF593330.1 | - | 4 | 382 | |||