Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK8 SIRNA Assays ›
Gene Symbol:
MAPK8
Gene Name:
mitogen-activated protein kinase 8
Gene Aliases:
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c
Chromosome Location:
Chr.10: 48306639 - 48439360 on Build GRCh38
UniGene ID:
Hs.138211
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1320
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001278547.1
NP_001265476.1
2, 3
352
NM_001278548.1
NP_001265477.1
1, 2
169
NM_001323302.1
NP_001310231.1
4, 5
615
NM_001323320.1
NP_001310249.1
2, 3
352
NM_001323321.1
NP_001310250.1
2, 3
352
NM_001323322.1
NP_001310251.1
4, 5
615
NM_001323323.1
NP_001310252.1
3, 4
421
NM_001323324.1
NP_001310253.1
2, 3
352
NM_001323325.1
NP_001310254.1
3, 4
546
NM_001323326.1
NP_001310255.1
3, 4
421
NM_001323327.1
NP_001310256.1
3, 4
546
NM_001323328.1
NP_001310257.1
3, 4
421
NM_001323329.1
NP_001310258.1
2, 3
352
NM_001323330.1
NP_001310259.1
3, 4
546
NM_001323331.1
NP_001310260.1
3, 4
546
NM_139046.3
NP_620634.1
4, 5
615
NM_139049.3
NP_620637.1
4, 5
615
NR_136583.1
2, 3
352
NR_136584.1
2, 3
352
NR_136585.1
2, 3
352
GenBank mRNA
AB451231.1
-
1, 2
120
AB451271.1
-
1, 2
120
AB451354.1
-
1, 2
120
AB451398.1
-
1, 2
120
AK292523.1
-
2, 3
248
BC130570.1
-
1, 2
182
40146601
BC130572.1
-
1, 2
182
40146603
BC144063.1
-
1, 2
182
9052583
DA159335.1
-
2, 3
352
DQ234352.1
-
1, 2
120
L26318.1
-
1, 2
138
U34822.1
-
1, 2
132
U35004.1
-
1, 2
138
U35005.1
-
1, 2
133
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
response to stress
JNK cascade
JUN phosphorylation
response to UV
positive regulation of gene expression
regulation of macroautophagy
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
regulation of histone deacetylation
positive regulation of cyclase activity
negative regulation of protein binding
regulation of protein localization
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
positive regulation of apoptotic process
negative regulation of apoptotic process
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
positive regulation of protein metabolic process
cellular response to lipopolysaccharide
cellular response to mechanical stimulus
positive regulation of deacetylase activity
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
protein serine/threonine kinase activity
JUN kinase activity
protein binding
ATP binding
kinase activity
enzyme binding
histone deacetylase regulator activity
histone deacetylase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0