Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCKL1 SIRNA Assays ›
Gene Symbol:
UCKL1
Gene Name:
uridine-cytidine kinase 1 like 1
Gene Aliases:
UCK1L, URKL1
Chromosome Location:
Chr.20: 63939829 - 63959435 on Build GRCh38
UniGene ID:
Hs.504998
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132133
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCKL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001193379.1
NP_001180308.1
14
1853
NM_017859.3
NP_060329.2
14
1625
NR_126526.1
14
1602
XM_005260216.2
XP_005260273.1
14
1593
XM_006723806.2
XP_006723869.1
14
1599
XM_006723807.2
XP_006723870.1
14
1596
XM_006723808.1
XP_006723871.1
14
1651
XM_006723809.1
XP_006723872.1
14
1631
XM_006723810.1
XP_006723873.1
14
1628
XM_006723811.3
XP_006723874.1
14
3620
XM_011528868.2
XP_011527170.1
14
3623
XM_011528869.1
XP_011527171.1
15
4894
XM_011528870.1
XP_011527172.1
14
1613
XM_011528871.1
XP_011527173.1
14
1640
XM_011528872.1
XP_011527174.1
15
4883
XM_017027896.1
XP_016883385.1
14
1628
XM_017027898.1
XP_016883387.1
14
1625
XM_017027899.1
XP_016883388.1
14
1622
XM_017027900.1
XP_016883389.1
14
2474
XM_017027901.1
XP_016883390.1
14
2469
XM_017027902.1
XP_016883391.1
15
1831
XM_017027903.1
XP_016883392.1
14
1634
XM_017027904.1
XP_016883393.1
14
4555
XM_017027905.1
XP_016883394.1
14
4552
XM_017027906.1
XP_016883395.1
14
4549
XM_017027907.1
XP_016883396.1
10
1351
XM_017027908.1
XP_016883397.1
11
1254
GenBank mRNA
AJ605558.1
-
14
1571
AK000524.1
-
14
1594
AK222559.1
-
14
1596
AK223220.1
-
14
1594
AK303688.1
-
14
1805
BC033078.1
-
12
1612
4810505
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
pyrimidine nucleobase metabolic process
viral process
phosphorylation
pyrimidine nucleoside salvage
UMP salvage
CTP salvage
uridine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0