Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R3 SIRNA Assays ›
Gene Symbol:
PPP6R3
Gene Name:
protein phosphatase 6 regulatory subunit 3
Gene Aliases:
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location:
Chr.11: 68460712 - 68615334 on Build GRCh38
UniGene ID:
Hs.503022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132428
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164160.1
NP_001157632.1
8
1065
NM_001164161.1
NP_001157633.1
8
1065
NM_001164162.1
NP_001157634.1
8
1065
NM_001164163.1
NP_001157635.1
8
1065
NM_001164164.1
NP_001157636.1
9
1170
NM_018312.4
NP_060782.2
9
1170
XM_006718608.3
XP_006718671.1
9
1181
XM_006718612.3
XP_006718675.1
9
1181
XM_006718613.2
XP_006718676.1
9
1181
XM_006718614.2
XP_006718677.1
9
1181
XM_006718618.3
XP_006718681.1
9
1181
XM_006718621.2
XP_006718684.1
9
1181
XM_006718622.3
XP_006718685.1
9
1181
XM_006718623.3
XP_006718686.1
9
1164
XM_006718624.2
XP_006718687.1
9
1181
XM_006718627.3
XP_006718690.1
9
1181
XM_011545135.1
XP_011543437.1
9
1181
XM_011545136.2
XP_011543438.1
8
1064
XM_011545137.2
XP_011543439.1
9
1164
XM_011545138.2
XP_011543440.1
10
1382
XM_011545139.1
XP_011543441.1
9
1181
XM_011545140.2
XP_011543442.1
9
1181
XM_011545141.1
XP_011543443.1
9
1181
XM_011545142.2
XP_011543444.1
9
1181
XM_011545143.1
XP_011543445.1
9
1181
XM_011545145.2
XP_011543447.1
9
1181
XM_017017962.1
XP_016873451.1
8
1019
XM_017017963.1
XP_016873452.1
9
1105
XM_017017964.1
XP_016873453.1
9
1181
XM_017017965.1
XP_016873454.1
8
1064
XM_017017966.1
XP_016873455.1
9
1267
XM_017017967.1
XP_016873456.1
9
1181
XM_017017968.1
XP_016873457.1
8
1064
XM_017017969.1
XP_016873458.1
9
1181
XM_017017970.1
XP_016873459.1
8
1064
XM_017017971.1
XP_016873460.1
9
1181
XM_017017972.1
XP_016873461.1
8
1037
XM_017017973.1
XP_016873462.1
9
1164
XM_017017974.1
XP_016873463.1
9
1181
XM_017017975.1
XP_016873464.1
9
1181
XM_017017976.1
XP_016873465.1
8
1064
XM_017017977.1
XP_016873466.1
8
952
XM_017017978.1
XP_016873467.1
7
803
XM_017017979.1
XP_016873468.1
7
803
XM_017017980.1
XP_016873469.1
7
803
XM_017017981.1
XP_016873470.1
7
803
XM_017017982.1
XP_016873471.1
7
803
XM_017017983.1
XP_016873472.1
8
1202
XM_017017984.1
XP_016873473.1
8
1132
GenBank mRNA
AB046778.2
-
8
1013
AF264779.1
-
8
1075
AF264780.1
-
8
1075
AK057250.1
-
2
138
AK315136.1
-
8
1034
BC105933.1
-
7
933
40029064
BC105934.1
-
7
933
40029058
BC107599.1
-
7
933
40029065
CR749815.1
-
8
1028
CR933658.1
-
8
1028
CR936718.1
-
8
1033
DQ111954.2
-
6
798
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of phosphoprotein phosphatase activity
COPII vesicle coating
protein binding
protein phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0