Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA1217 SIRNA Assays ›
Gene Symbol:
KIAA1217
Gene Name:
KIAA1217
Gene Aliases:
SKT
Chromosome Location:
Chr.10: 23694746 - 24547848 on Build GRCh38
UniGene ID:
Hs.445885
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132876
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA1217
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098500.2
NP_001091970.1
7
1944
NM_001282767.1
NP_001269696.1
6
1778
NM_001282768.1
NP_001269697.1
6
1778
NM_001282769.1
NP_001269698.1
2
861
NM_001282770.1
NP_001269699.1
2
815
NM_001321681.1
NP_001308610.1
2
861
NM_019590.4
NP_062536.2
6
1778
XM_005252516.3
XP_005252573.1
2
789
XM_011519552.2
XP_011517854.2
7
1833
XM_011519555.2
XP_011517857.2
6
1751
XM_011519558.2
XP_011517860.2
7
1835
XM_011519559.2
XP_011517861.2
6
1525
XM_011519562.2
XP_011517864.2
7
1833
XM_011519564.2
XP_011517866.1
6
1505
XM_011519565.2
XP_011517867.2
6
1524
XM_011519566.2
XP_011517868.2
6
1752
XM_017016416.1
XP_016871905.1
7
1859
XM_017016417.1
XP_016871906.1
6
1778
XM_017016418.1
XP_016871907.1
5
2495
XM_017016419.1
XP_016871908.1
5
2495
XM_017016420.1
XP_016871909.1
5
1242
XM_017016421.1
XP_016871910.1
7
1452
XM_017016422.1
XP_016871911.1
5
1242
XM_017016423.1
XP_016871912.1
2
789
XM_017016424.1
XP_016871913.1
2
861
XM_017016425.1
XP_016871914.1
7
1835
XM_017016426.1
XP_016871915.1
7
1839
XM_017016427.1
XP_016871916.1
7
1605
XM_017016428.1
XP_016871917.1
4
1021
XM_017016429.1
XP_016871918.1
8
1993
GenBank mRNA
AB033043.2
-
7
1944
AB291616.1
-
5
1305
AK091142.1
-
6
1778
AK096446.1
-
5
1166
AK300422.1
-
2
800
AK303905.1
-
5
1290
AL833280.1
-
6
1405
BC018764.1
-
2
629
BC098577.1
-
2
769
BC136521.1
-
6
1396
9020511
BC144227.1
-
6
1396
9052747
BX640796.1
-
2
790
BX648451.1
-
2
790
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
embryonic skeletal system development
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-bz4hs:80/100.66.77.47:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0