Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13310
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
2
431
NM_001220474.2
NP_001207403.1
2
431
NM_001220475.2
NP_001207404.1
2
243
NM_001220476.2
NP_001207405.1
2
243
NM_003625.4
NP_003616.2
3
294
XM_011538906.2
XP_011537208.1
2
425
XM_011538907.2
XP_011537209.1
2
425
XM_017020079.1
XP_016875568.1
3
278
XM_017020080.1
XP_016875569.1
2
425
XM_017020081.1
XP_016875570.1
2
425
XM_017020082.1
XP_016875571.1
3
278
XM_017020083.1
XP_016875572.1
2
425
XM_017020084.1
XP_016875573.1
2
425
XM_017020085.1
XP_016875574.1
2
425
XM_017020086.1
XP_016875575.1
3
278
XM_017020087.1
XP_016875576.1
2
412
XM_017020088.1
XP_016875577.1
3
712
XM_017020089.1
XP_016875578.1
2
425
XM_017020090.1
XP_016875579.1
3
278
XM_017020091.1
XP_016875580.1
2
425
XM_017020092.1
XP_016875581.1
2
425
XM_017020093.1
XP_016875582.1
2
425
XM_017020094.1
XP_016875583.1
3
278
XM_017020095.1
XP_016875584.1
2
425
XM_017020096.1
XP_016875585.1
2
425
XM_017020097.1
XP_016875586.1
3
278
XM_017020098.1
XP_016875587.1
3
278
XM_017020099.1
XP_016875588.1
2
425
XM_017020100.1
XP_016875589.1
2
425
XM_017020101.1
XP_016875590.1
3
278
XM_017020102.1
XP_016875591.1
2
412
XM_017020103.1
XP_016875592.1
2
425
XM_017020104.1
XP_016875593.1
2
425
XM_017020105.1
XP_016875594.1
2
425
XM_017020106.1
XP_016875595.1
2
425
XM_017020107.1
XP_016875596.1
3
278
XM_017020108.1
XP_016875597.1
3
278
XM_017020109.1
XP_016875598.1
2
425
XM_017020110.1
XP_016875599.1
3
712
XM_017020111.1
XP_016875600.1
2
425
GenBank mRNA
AB210009.1
-
2
429
AF034799.1
-
3
302
AK126971.1
-
2
391
AK307524.1
-
3
278
BC104912.1
-
2
227
8143915
BC143485.1
-
2
227
9051993
DA499362.1
-
2
432
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline