Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADAM22 SIRNA Assays ›
Gene Symbol:
ADAM22
Gene Name:
ADAM metallopeptidase domain 22
Gene Aliases:
ADAM 22, MDC2
Chromosome Location:
Chr.7: 87934143 - 88202889 on Build GRCh38
UniGene ID:
Hs.256398
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133416
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADAM22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324417.1
NP_001311346.1
17
1739
NM_001324418.1
NP_001311347.1
17
1742
NM_001324419.1
NP_001311348.1
17
1739
NM_001324420.1
NP_001311349.1
17
1742
NM_001324421.1
NP_001311350.1
17
1742
NM_004194.4
NP_004185.1
17
1742
NM_016351.5
NP_057435.2
17
1742
NM_021721.4
NP_068367.1
17
1742
NM_021722.5
NP_068368.2
17
1742
NM_021723.4
NP_068369.1
17
1742
XM_005250445.3
XP_005250502.1
16
1590
XM_006716028.3
XP_006716091.1
16
1590
XM_006716029.3
XP_006716092.1
16
1590
XM_011516318.1
XP_011514620.1
17
1634
XM_011516319.2
XP_011514621.1
17
1718
XM_011516320.2
XP_011514622.1
16
1590
XM_011516321.2
XP_011514623.1
16
1590
XM_011516322.1
XP_011514624.1
17
1634
XM_011516323.1
XP_011514625.1
17
1634
XM_011516324.1
XP_011514626.1
17
1306
XM_017012329.1
XP_016867818.1
18
1685
XM_017012330.1
XP_016867819.1
18
1685
XM_017012331.1
XP_016867820.1
17
1715
XM_017012332.1
XP_016867821.1
17
1634
XM_017012333.1
XP_016867822.1
17
1634
XM_017012334.1
XP_016867823.1
18
1685
XM_017012335.1
XP_016867824.1
18
1685
XM_017012336.1
XP_016867825.1
17
1634
XM_017012337.1
XP_016867826.1
18
1685
XM_017012338.1
XP_016867827.1
17
1713
XM_017012339.1
XP_016867828.1
18
1685
XM_017012340.1
XP_016867829.1
6
365
XM_017012341.1
XP_016867830.1
16
1362
GenBank mRNA
AB009671.1
-
17
1540
AF073291.2
-
17
1465
AF155381.1
-
17
1465
AF155382.1
-
17
1465
AF158637.1
-
16
1302
AK125432.1
-
16
1605
AK291010.1
-
17
1589
BC125118.1
-
17
1496
40118904
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
cell adhesion
negative regulation of cell adhesion
central nervous system development
adult locomotory behavior
myelination in peripheral nervous system
metalloendopeptidase activity
integrin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline