Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CIZ1 SIRNA Assays ›
Gene Symbol:
CIZ1
Gene Name:
CDKN1A interacting zinc finger protein 1
Gene Aliases:
LSFR1, NP94, ZNF356
Chromosome Location:
Chr.9: 128166065 - 128204383 on Build GRCh38
UniGene ID:
Hs.212395
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133814
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CIZ1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001131015.1
NP_001124487.1
9
1474
NM_001131016.1
NP_001124488.1
8
1602
NM_001131017.1
NP_001124489.1
9
1419
NM_001131018.1
NP_001124490.1
8
1362
NM_001257975.1
NP_001244904.1
8
1694
NM_001257976.1
NP_001244905.1
7
1486
NM_012127.2
NP_036259.2
8
1642
XM_005251888.3
XP_005251945.2
8
1473
XM_005251891.3
XP_005251948.2
9
1305
XM_005251892.3
XP_005251949.2
9
1290
XM_005251893.4
XP_005251950.4
8
1228
XM_006717039.3
XP_006717102.2
7
1381
XM_011518481.1
XP_011516783.1
8
1458
XM_011518482.2
XP_011516784.2
7
1396
XM_011518483.1
XP_011516785.1
7
1513
XM_017014594.1
XP_016870083.1
8
1926
XM_017014595.1
XP_016870084.1
8
1570
XM_017014596.1
XP_016870085.1
9
1417
GenBank mRNA
AB030835.1
-
8
1555
AF159025.1
-
7
1434
AF234161.1
-
6
1205
AK023978.1
-
9
1474
AK292713.1
-
8
1593
AK296363.1
-
8
1695
AK303288.1
-
9
1402
AK303636.1
-
7
1469
AK316393.1
-
6
1198
BC004119.2
-
9
1259
3507940
BC021163.2
-
8
1341
3633796
EF441870.1
-
1
148
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleic acid binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0