Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUP62 SIRNA Assays ›
          Gene Symbol:
          NUP62
          Gene Name:
          nucleoporin 62
          Gene Aliases:
          IBSN, SNDI, p62
          Chromosome Location:
          Chr.19: 49906825 - 49929731 on Build GRCh38
          UniGene ID:
          Hs.574492
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 133922

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUP62
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193357.1 NP_001180286.1 2 2502
          NM_012346.4 NP_036478.2 2 2543
          NM_016553.4 NP_057637.2 3 2904
          NM_153718.3 NP_714940.1 2 2582
          NM_153719.3 NP_714941.1 3 2737
          GenBank mRNA AF318357.1 - 3 1284
          AL162061.1 - 2 2503

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          RNA export from nucleus
          mRNA export from nucleus
          tRNA export from nucleus
          protein import into nucleus
          mitotic nuclear division
          mitotic nuclear envelope disassembly
          cell surface receptor signaling pathway
          spermatogenesis
          cell aging
          cell death
          negative regulation of cell proliferation
          hormone-mediated signaling pathway
          regulation of signal transduction
          regulation of glucose transport
          viral process
          protein sumoylation
          viral transcription
          gene silencing by RNA
          negative regulation of epidermal growth factor receptor signaling pathway
          regulation of protein import into nucleus
          negative regulation of apoptotic process
          negative regulation of programmed cell death
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of MAP kinase activity
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of transcription, DNA-templated
          regulation of Ras protein signal transduction
          negative regulation of Ras protein signal transduction
          protein heterotrimerization
          ribonucleoprotein complex export from nucleus
          intracellular transport of virus
          regulation of cellular response to heat
          chromatin binding
          nucleocytoplasmic transporter activity
          protein binding
          phospholipid binding
          structural constituent of nuclear pore
          kinesin binding
          receptor signaling complex scaffold activity
          Hsp70 protein binding
          SH2 domain binding
          ubiquitin binding
          thyroid hormone receptor binding
          PTB domain binding
          Hsp90 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0