Search
Search

SNX12| Gene Symbol: SNX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256185.1 | NP_001243114.1 | 3 | 360 | ||
| NM_001256186.1 | NP_001243115.1 | 2 | 324 | |||
| NM_001256187.1 | NP_001243116.1 | 2 | 336 | |||
| NM_001256188.1 | NP_001243117.1 | 2 | 324 | |||
| NM_013346.3 | NP_037478.2 | 2 | 336 | |||
| GenBank mRNA | AF171229.1 | - | 2 | 213 | ||
| AK312977.1 | - | 2 | 284 | |||
| BC020559.1 | - | 2 | 228 | |||
| BC080535.1 | - | 2 | 211 | |||
| BC103847.2 | - | 2 | 226 | 40002749 | ||
| BC103848.1 | - | 2 | 226 | 40002752 | ||
| BM805683.1 | - | 383 | ||||
| BT007203.1 | - | 2 | 194 | |||
| DA439434.1 | - | 2 | 272 | |||
| DA736880.1 | - | 3 | 360 | |||
| DA847821.1 | - | 2 | 277 | |||
| DB251077.1 | - | 2 | 292 | |||
| DB482683.1 | - | 2 | 336 | |||