Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIPA1L1 SIRNA Assays ›
Gene Symbol:
SIPA1L1
Gene Name:
signal induced proliferation associated 1 like 1
Gene Aliases:
E6TP1
Chromosome Location:
Chr.14: 71320418 - 71741229 on Build GRCh38
UniGene ID:
Hs.654657
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
134562
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIPA1L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284245.1
NP_001271174.1
10
3526
NM_001284246.1
NP_001271175.1
10
4115
NM_001284247.1
NP_001271176.1
9
3467
NM_015556.2
NP_056371.1
10
3526
XM_005267516.4
XP_005267573.1
10
4115
XM_006720111.2
XP_006720174.1
12
5405
XM_011536630.1
XP_011534932.1
14
3877
XM_011536631.1
XP_011534933.1
14
4288
XM_011536632.1
XP_011534934.1
13
3797
XM_011536635.2
XP_011534937.1
13
3944
XM_011536638.2
XP_011534940.1
9
2600
XM_017021159.1
XP_016876648.1
16
4034
XM_017021160.1
XP_016876649.1
14
3956
XM_017021161.1
XP_016876650.1
15
3909
XM_017021162.1
XP_016876651.1
17
4091
XM_017021163.1
XP_016876652.1
12
3758
XM_017021164.1
XP_016876653.1
12
3765
XM_017021165.1
XP_016876654.1
14
3834
XM_017021166.1
XP_016876655.1
15
3967
XM_017021167.1
XP_016876656.1
14
3842
XM_017021168.1
XP_016876657.1
14
3816
XM_017021169.1
XP_016876658.1
13
3969
XM_017021170.1
XP_016876659.1
13
4327
XM_017021171.1
XP_016876660.1
14
3900
XM_017021172.1
XP_016876661.1
13
3775
XM_017021173.1
XP_016876662.1
12
3902
XM_017021174.1
XP_016876663.1
12
4142
XM_017021175.1
XP_016876664.1
12
4260
XM_017021176.1
XP_016876665.1
17
4087
XM_017021177.1
XP_016876666.1
17
4076
XM_017021178.1
XP_016876667.1
15
3914
XM_017021179.1
XP_016876668.1
14
3817
XM_017021180.1
XP_016876669.1
18
4129
XM_017021181.1
XP_016876670.1
16
4034
XM_017021182.1
XP_016876671.1
15
3967
XM_017021183.1
XP_016876672.1
13
3775
XM_017021184.1
XP_016876673.1
17
4087
XM_017021185.1
XP_016876674.1
10
4115
XM_017021186.1
XP_016876675.1
16
4034
XM_017021187.1
XP_016876676.1
14
3900
XM_017021188.1
XP_016876677.1
13
3775
XM_017021189.1
XP_016876678.1
15
3884
XM_017021190.1
XP_016876679.1
16
4034
XM_017021191.1
XP_016876680.1
14
3842
XM_017021192.1
XP_016876681.1
13
3775
XM_017021193.1
XP_016876682.1
13
3773
XM_017021194.1
XP_016876683.1
16
4034
XM_017021195.1
XP_016876684.1
16
4034
XM_017021196.1
XP_016876685.1
16
4034
XM_017021197.1
XP_016876686.1
15
2234
XM_017021198.1
XP_016876687.1
12
1975
XM_017021199.1
XP_016876688.1
16
2276
XM_017021200.1
XP_016876689.1
13
2042
GenBank mRNA
AB007900.2
-
9
3250
AF090989.1
-
10
3513
AF090990.1
-
10
3513
AK122930.1
-
10
4115
AK302650.1
-
9
1765
BC146777.1
-
9
3250
9007226
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
actin cytoskeleton reorganization
regulation of GTPase activity
ephrin receptor signaling pathway
regulation of synaptic plasticity
regulation of axonogenesis
regulation of small GTPase mediated signal transduction
regulation of dendritic spine morphogenesis
activation of GTPase activity
molecular_function
GTPase activator activity
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0