Search
Search

TES| Gene Symbol: TES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015641.3 | NP_056456.1 | 7 | 1514 | ||
| NM_152829.2 | NP_690042.1 | 7 | 1348 | |||
| XM_005250258.1 | XP_005250315.1 | 6 | 1382 | |||
| GenBank mRNA | AF245356.1 | - | 7 | 1440 | ||
| AF245357.1 | - | 7 | 1318 | |||
| AJ250865.1 | - | 7 | 1427 | |||
| AK021575.1 | - | 7 | 1458 | |||
| AK222840.1 | - | 7 | 1444 | |||
| AK291802.1 | - | 7 | 1329 | |||
| AK300546.1 | - | 5 | 869 | |||
| AL050162.1 | - | 3 | 446 | |||
| BC001451.1 | - | 7 | 1480 | 3138867 | ||