Search
Search

SH3YL1| Gene Symbol: SH3YL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159597.2 | NP_001153069.1 | 7 | 773 | ||
| NM_001282682.1 | NP_001269611.1 | 9 | 878 | |||
| NM_001282687.1 | NP_001269616.1 | 9 | 927 | |||
| NM_015677.3 | NP_056492.2 | 7 | 773 | |||
| NR_104223.1 | 9 | 2332 | ||||
| NR_104225.1 | 7 | 968 | ||||
| NR_104226.1 | 9 | 1493 | ||||
| NR_104227.1 | 9 | 1839 | ||||
| GenBank mRNA | AK026507.1 | - | 7 | 708 | ||
| AK026586.1 | - | 7 | 968 | |||
| AK055801.1 | - | 7 | 1432 | |||
| AK096440.1 | - | 9 | 1417 | |||
| AK123829.1 | - | 10 | 2571 | |||
| AK292312.1 | - | 9 | 822 | |||
| AL050373.1 | - | 9 | 940 | |||
| BC008374.1 | - | 7 | 682 | 3996066 | ||
| BC008375.1 | - | 7 | 700 | 3996120 | ||
| BC043234.1 | - | 9 | 2337 | |||
| CR457361.1 | - | 6 | 660 | |||