Search
Search

LARS| Gene Symbol: LARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317964.1 | NP_001304893.1 | 6, 7 | 869 | ||
| NM_001317965.1 | NP_001304894.1 | 6, 7 | 888 | |||
| NM_016460.3 | NP_057544.2 | 6, 7 | 926 | |||
| NM_020117.10 | NP_064502.9 | 7, 8 | 1007 | |||
| XM_011537656.2 | XP_011535958.1 | 7, 8 | 873 | |||
| GenBank mRNA | AB037773.1 | - | 7, 8 | 880 | ||
| AK025441.1 | - | 7, 8 | 843 | |||
| AK225372.1 | - | 6, 7 | 705 | |||
| AK293750.1 | - | 6, 7 | 756 | |||
| AK295874.1 | - | 6, 7 | 724 | |||
| AK304134.1 | - | 6, 7 | 685 | |||
| AK315178.1 | - | 7, 8 | 856 | |||
| BC150213.1 | - | 7, 8 | 858 | 40148958 | ||
| BC151214.1 | - | 7, 8 | 808 | 40148007 | ||
| BC152422.1 | - | 7, 8 | 880 | 8862534 | ||
| D84223.1 | - | 7, 8 | 785 | |||