Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TPK1 SIRNA Assays ›
          Gene Symbol:
          TPK1
          Gene Name:
          thiamin pyrophosphokinase 1
          Gene Aliases:
          HTPK1, PP20, THMD5
          Chromosome Location:
          Chr.7: 144451941 - 144836424 on Build GRCh38
          UniGene ID:
          Hs.660232
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 135309

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TPK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042482.1 NP_001035947.1 5, 6 369
          NM_022445.3 NP_071890.2 5, 6 369
          XM_005249970.1 XP_005250027.1 5, 6 347
          XM_011516031.1 XP_011514333.1 6, 7 448
          XM_011516032.2 XP_011514334.1 6, 7 584
          XM_011516033.2 XP_011514335.1 6, 7 454
          XM_011516034.2 XP_011514336.1 6, 7 511
          XM_011516035.2 XP_011514337.1 6, 7 448
          XM_011516037.2 XP_011514339.1 5, 6 695
          XM_011516039.2 XP_011514341.1 5, 6 630
          XM_011516040.2 XP_011514342.1 6, 7 448
          XM_011516041.2 XP_011514343.1 4, 5 598
          XM_011516043.1 XP_011514345.1 6, 7 448
          XM_011516044.1 XP_011514346.1 3, 4 272
          XM_011516046.1 XP_011514348.1 6, 7 448
          XM_011516047.2 XP_011514349.1 2, 3 1119
          XM_011516048.1 XP_011514350.1 1, 2 85
          XM_011516050.1 XP_011514352.1 3, 4 178
          XM_017011969.1 XP_016867458.1 6, 7 448
          XM_017011970.1 XP_016867459.1 5, 6 422
          XM_017011971.1 XP_016867460.1 5, 6 372
          XM_017011972.1 XP_016867461.1 6, 7 448
          XM_017011973.1 XP_016867462.1 5, 6 514
          XM_017011974.1 XP_016867463.1 5, 6 359
          XM_017011975.1 XP_016867464.1 5, 6 310
          XM_017011976.1 XP_016867465.1 5, 6 369
          XM_017011977.1 XP_016867466.1 6, 7 462
          XM_017011978.1 XP_016867467.1 5, 6 421
          XM_017011979.1 XP_016867468.1 5, 6 501
          XM_017011982.1 XP_016867471.1 3, 4 268
          XM_017011983.1 XP_016867472.1 3, 4 212
          XM_017011984.1 XP_016867473.1 3, 4 271
          XM_017011985.1 XP_016867474.1 3, 4 227
          XM_017011986.1 XP_016867475.1 3, 4 233
          GenBank mRNA AB028138.1 - 4, 5 284
          AF297710.1 - 5, 6 356
          AK026374.1 - 3, 4 176
          AK091736.1 - 5, 6 308
          AK128661.1 - 4, 5 322
          AK289652.1 - 5, 6 344
          AK308693.1 - 5, 6 369
          AY206415.1 - 5, 6 351
          BC040555.1 - 6, 7 517 5269277
          BC068460.1 - 5, 6 348 5288626
          BI770816.1 - 4, 5 283
          CR749248.1 - 3, 4 356
          DA135998.1 - 5, 6 369

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          thiamine metabolic process
          thiamine diphosphate biosynthetic process
          phosphorylation
          thiamine-containing compound metabolic process
          thiamine diphosphokinase activity
          ATP binding
          kinase activity
          thiamine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0